missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Bioanalysis™ Nucleospin 96 Tissue

Product Code. 12778552
Click to view available options
:
24x 96-well plates for the isolation of genomic DNA from tissue, buffer T1, BQ1, B5, BW, BE Proteinase K, NucleoSpin tissue binding plates, protocol Macherey-Nagel
This item is not returnable. View return policy

Product Code. 12778552

Brand: Macherey Nagel Bioanalysis™ 740741.24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
NucleoSpin™ 96 Tissue is designed for the rapid simultaneous preparation of highly pure genomic DNA from animal and human tissue, eg. mouse and rat tails.
From a 1.2cm mouse tail tip section (age of mice 4 to 6 weeks), up to 30μg to 40μg of pure genomic DNA can be prepared (typical yields 15μg to 25μg). The obtained DNA can be used directly for PCR*, Southern blotting or any kind of enzymatic reaction.NucleoSpin™ Tissue binding plates can be used under vacuum or centrifugation in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. Up to 192 samples (two plates) can be processed in less than one hour (after lysis). With the NucleoSpin™ 96 Tissue method, lysis is achieved by incubation of the samples in a solution containing detergents and proteinase K at 56°C. Appropriate conditions for binding of DNA to the silica membranes of the NucleoSpin™ Tissue binding plate are created by addition of large amounts of chaotropic ions plus ethanol to the lysate. The binding process is reversible and specific to nucleic acids. Contaminants are removed by washing with two different buffers. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer.Kit components: NucleoSpin™ Tissue binding plates, buffers, proteinase K, wash plates, square-well blocks, round-well blocks, racks with tube strips, cap strips, self-adhesing foil. For further details on this product please contact Customer Services.
  • Silica membrane technology
  • Format: 96 well plates
  • Preparation time: 1hr (after lysis)
  • Sample material: ≤20mg tissue, 106 cultured or bacteria
  • Yield: 15μg to 25μg
  • Elution volume: 100μL to 200μL
  • Fragment size: 300bp to 30kb
  • No use of organic solvents
For manual or automated isolation of DNA from human and animal tissue, cultured cells or bacteria.

Specifications

Format Standard
No. of Samples 96
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride, 2-propanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.