missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoSpin™ 96 Tissue Core Kit

Allround kit for purification of DNA from a broad range of samples in a proven 96‐well plate format.

Brand:  Macherey-Nagel Bioanalysis™ 740454.4

Product Code. 12778422

  • 960.00 € / 384 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sample Material: Animal tissue, Bacteria, Cells, Human tissue
  • Sample Amount: < 20 mg human / animal tissue, < 106 cells
  • Fragment Size: 300 bp to 50 kbp
  • Preparation Time: 60 min/plate (excludes lysis)
Specifications

Specifications

18°C to 25°C
Silica Membrane Technology
384 Preps
96-well Plate
Isolation of DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers