missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Blood, Mini kit for DNA from blood
Description
Features:
- For rapid purification of high-quality DNA from blood
- Ready-to-use DNA in less than 30 min
- Complete removal of PCR inhibitors
- Suitable for whole blood, treated with citrate, EDTA, heparin, CPDA
- Consistently high DNA yields
- Suitable for isolation of viral DNA
Applications:
- Genomic, bacterial, viral DNA isolation
- DNA from whole blood (human or animal blood, fresh or frozen)
- DNA from whole blood treated with citrate, EDTA, heparin, CPDA
- DNA from serum, plasma, buffy coat, platelets, body fluids (e.g. amniotic fluid)
- DNA from cultured cells
- DNA suitable for (PCR, Southern blotting, enzymatic reactions, etc.)
Specifications
Specifications
| Detection Method | Silica-membrane Technology |
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane Technology |
| For Use With (Equipment) | PCR, Southern Blotting, Enzymatic Reactions |
| For Use With (Application) | For Genomic, Bacterial, Viral DNA Isolation |
| Includes | 2mL Collection Tubes, Buffers, Proteinase K |
| No. of Reactions | 10 |
| Product Type | NucleoSpin™ Blood Column |
| Quantity | 1 Each |
| Test Time | 30min. |
| Show More |
Safety and Handling
Product Identifier
- NucleoSpin Blood
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash with water thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: guanidine hydrochloride / 2-propanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?