missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ DNA FFPE XS, Micro kit for DNA from FFPE

Product Code. 11932502
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 11932502

Brand: Macherey-Nagel™ 740980.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Improved DNA quality from formalin-fixed, paraffin-embedded samples - no xylene needed.

Collection tubes (2mL) paraffin desolver, buffers, Proteinase K 250 preps Macherey Nagel

Specifications

Buffer Yes
Final Product Type NucleoSpin DNA FFPE XS
No. of Reactions 10
Product Type NucleoSpin DNA FFPE XS
Sample Volume ≤7 sections (10 μm) of 250 mm2 total area, <15 mg paraffin
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin DNA FFPE XS (10)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : paraffin disolver, proteinase k
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.