missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up, kit for gel extraction or PCR clean up

Product Code. 12303368
Change view
Click to view available options
No. of Reactions:
10 Preps
240 Preps
50 Preps
Unit Size:
10 reactions
240 preps
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12303368 50 Preps 50 reactions
11992462 10 Preps 10 reactions
15116456 240 Preps 240 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12303368 Supplier Macherey-Nagel™ Supplier No. 740609.50
  • ONLINE EXCLUSIVE PRICE
    173.00 €
    121.80 € / 50 reactions
    Save 51.20 €
    29% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clean up PCR reactions and gel extractions with a two-in-one approach. Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Kit is optimized for recovery and elution volume when purifying DNA fragments via both applications.

NucleoSpin™ Gel and PCR Clean-up is designed for fast purification of PCR products, such as DNA from enzymatic reactions, as well as the extraction of DNA fragments from TAE or TBE agarose gels.

  • Two applications in one kit—One buffer to process both sample types, with only one loading step
  • High recovery for fragments down to 50 bp—Average recovery from 60–90%
  • Minimized elution volume of 15μL—Highly concentrated DNA
  • Binding buffer with pH indicator—Ensures optimal binding conditions
  • Separate buffers for single stranded DNA and RNA or for SDS-containing samples available separately
  • Suitable for all kinds of agarose gels and a variety of buffer systems (e.g. TAE, TBE)
  • Complete removal of contaminations: nucleotides, primers, enzymes, mineral oil, PCR additives, detergents, dyes
  • Applications: purification of PCR products, extraction of DNA from agarose or polyacrylamide gels, and downstream applications (cloning, sequencing, PCR, restriction analysis)

Kit contents: NucleoSpin™ Gel and PCR Clean-up columns, collection tubes (2mL), binding buffer with pH indicator, wash buffer and elution buffer

Specifications

Color pH Indicator:
Yellow = correct conditions
Green = pH slightly high (adjustment recommended)
Blue = pH too high (adjustment necessary)
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Efficiency Typical recovery: 70–95%
Elution Volume Elution volume: 15–30μL
For Use With (Equipment) TAE or TBE agarose gels, PCR
Includes Binding Buffer NTI (40mL), Wash Buffer NT3 Concentrate (25mL), Elution Buffer NE (13mL), NucleoSpin Gel and PCR Clean-up Columns (50), 2mL Collection Tubes (50)
No. of Reactions 50 Preps
Quantity 1 Kit
Purity A260/A280: 1.8–1.9
Sample Volume <200 μL PCR reaction mixture, <200 mg agarose gel
Size 50 bp to ∼20 kbp
Test Time 10 min/6 preps
Yield Up to ∼15μg DNA
Show More Show Less
compliance-icons
Product Identifier
  • NucleoSpin Gel and PCR Clean-up
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.