missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ miRNA, Mini kit for miRNA and RNA purification

Product Code. 11932512
Change view
Click to view available options
No. of Reactions:
10 Reactions
250 Reactions
50 Reactions
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
11932512 10 Reactions 10 reactions
12788622 250 Reactions 250 reactions
12798622 50 Reactions 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11932512 Supplier Macherey-Nagel™ Supplier No. 740971.10
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Isolate small and large RNA in parallel. NucleoSpin™ miRNA Kits work on a wide varitey of tissue and cell culture formats.

Features

  • Parallel isolation of small and large RNA
  • RNA purification fractionated by size:
  • Isolation of small RNA only (< 200 b),
  • Isolation of small RNA (< 200 b) and large RNA (> 200 b) in two separate fractions
  • Isolation of total RNA (small and large RNA in one fraction)
  • Additional isolation of total protein fraction ready to use for SDS-PAGE and Western blot analysis
  • Excellent RNA recovery and purity by chaotropic salt lysis without phenol/chloroform (patent pending)
  • NucleoSpin™ Filters for efficient sample homogenization
  • rDNase for efficient on-column removal of genomic DNA

Applications

  • Parallel isolation of small and large RNA from human / animal tissue and cultured cells, from plant tissue, and in combination with phenol / chloroform (e.g., TRIzol™) lysis
  • Purification of siRNA and large dsRNA from DICER reactions
  • Typical downstream applications: real-time RT-PCR, Northern blotting, chip hybridization

Specifications

Format Mini Spin Column
For Use With (Application) Purification of siRNA and large dsRNA from DICER reactions
Includes NucleoSpin™ RNA Columns, NucleoSpin™ miRNA Columns, NucleoSpin™ Protein Removal Columns, Collection Tubes (2 mL, 2 mL lid, 1.5 mL), NucleoSpin™ Filters, buffers, RNase-free rDNase
No. of Reactions 10 Reactions
Product Type NucleoSpin™ miRNA Kit
RNase Yes
Sample Size Small RNA: < 200 nt, large RNA: > 200 nt
Test Time <45 min./6 Reaction and <35 min./6 Reaction
Yield 10μg small RNA, 95μg large RNA
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin miRNA (10)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-dioxane, ethanol, guanidinium thiocyanate, 2-mercaptoethanol, rDNase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.