missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Plant, Mini kit for RNA from plant

Product Code. 12711021
Change view
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12711021 250 Preps 250 reactions
12979894 10 Preps 10 reactions
12721021 50 Preps 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12711021 Supplier Macherey-Nagel™ Supplier No. 740949.250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolates total RNA from all kinds of plant material and filamentous fungi without degradation of the RNA. Macherey-Nagel™ NucleoSpin™ RNA Plant Kit is a complete mini spin kit that includes RNA plant columns with collection tubes, filters, all needed buffers and RNase-free rDNase.

NucleoSpin™ RNA Plant Kit is designed for isolation of DNA-free, high-quality RNA from a wide variety of plant and fungal samples. Typical downstream applications include real-time RT-PCR, gene expression profiling, Northern blotting, primer extension, array technology, and RNase protection assays.

  • Two different lysis buffers included—Optimized lysis procedure for typical conditions and for peculiar metabolites
  • rDNase included for on-column digestion—Efficient removal of contaminating DNA
  • NucleoSpin™ Filters (shredders) included—Efficient sample homogenization and reduction of viscosity
  • Parallel purification of DNA possible—By using the NucleoSpin™ RNA/DNA buffer set

Kit Components: NucleoSpin™ RNA columns with 2 mL collection tubes, 2 mL and 1.5 mL collecting tubes, NucleoSpin™ filters, buffers (lysis, wash, desalting), RNase-free DNase I, reaction buffer for DNase, RNase-free water

  • Mini spin column with silica membrane technology
  • Yields 3 to 70μg RNA from 100μg plant material
  • Elution volume: 40 to 60μL
  • Fragment size: 200b–20kb
  • Binding capacity: 200μg
  • Preparation time: 30 min./6 preps
  • A260/A280: 1.9–2.1

Specifications

Format Mini Spin Column
For Use With (Equipment) Real-time RT-PCR
For Use With (Application) RNA purification from plant cells and tissue
Includes Lysis buffer RA1 (125mL), Lysis buffer RAP (125mL), Wash buffer RAW2 (80mL), Wash Buffer RA3 concentrate (3 x 25mL), Membrane desalting buffer (125mL), Reaction buffer for rDNase (30mL), RNase-free rDNase (5 size F vials), RNase-free water (60mL), NucleoSpin filters (250), NucleoSpin RNA Plan Columns with collection tubes (250), 2mL Collection tubes (750), 1.5mL Collection tubes (250)
No. of Reactions 250 Preps
Product Type NucleoSpin™ RNA Plant Column
Quantity 1 Each
Sample Size <100 mg
Yield 3–70μg
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Plant
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: ethanol (diluted < 20 %)(denatured with1%IPA/1%MEK, acc.2016/1867/EU); guanidinium thiocyanate; gua
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.