Learn More
Macherey-Nagel™ NucleoSpin™ cfDNA XS, Micro kit for cell-free DNA from plasma
Product Code. 12788562
Change view
Click to view available options
No. of Reactions:
10 Preps
250 Preps
Unit Size:
10 reactions
250 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12788562 250 Preps 250 reactions
12743570 10 Preps 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12788562 Supplier Macherey-Nagel™ Supplier No. 740900.250
  • ONLINE EXCLUSIVE PRICE
    895.00 €
    791.81 € / 250 reactions
    Save 103.19 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
XS spin kit for efficient isolation of cell-free DNA from small plasma samples.
Specifications
No. of Reactions 250 Preps
Product Type Plasma XS
Sample Volume <240 μL (up to 720 μL with multiple loading)