missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid Columns for plasmid DNA

Product Code. 15860928
Change view
Click to view available options
Quantity:
250 Pk.
50 Pk.
Unit Size:
Pack of 250
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860928 250 Pk. Pack of 250
15870928 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15860928 Supplier Macherey-Nagel™ Supplier No. 740588.250S
  • ONLINE EXCLUSIVE PRICE
    257.00 €
    215.25 € / Pack of 250
    Save 41.75 €
    16% Off
In Stock
Add to basket
Edge
Add to basket

Replacement columns for use with NucleoSpin™ Plasmid kits for rapid, small-scale preparation of high-purity, sequencing-grade plasmid DNA.

NucleoSpin™ Plasmid

  • For rapid small scale preparation high-purity plasmid DNA
  • Fast procedure – 6 Mini preps in 25 min
  • High capacity – up to 50 μg of plasmid DNA
  • Chaotropic salt wash buffer – extra pure DNA
  • Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
    • High-copy plasmids from E. coli
    • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
    • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
    • Supplementary protocol for M13 DNA availabl

Specifications

Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 50 μg
Elution Volume 50 μL
Final Product Type Sequencing-grade plasmid DNA
For Use With (Equipment) NucleoSpin™ Plasmid Kit
For Use With (Application) Preparation of high-purity plasmid DNA
Includes Columns only
Product Type NucleoSpin Plasmid lidded Column
Quantity 250 Pk.
Sample Volume 2 to 10 mL
Show More Show Less
Product Identifier
  • NucleoSpin Plasmid
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt/guanidine hydrochloride/2-pr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.