missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid (NoLid) Columns for plasmid DNA purification

Product Code. 15850918
Change view
Click to view available options
Quantity:
250 Pk.
50 Pk.
Unit Size:
Pack of 250
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15850918 250 Pk. Pack of 250
15860918 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15850918 Supplier Macherey-Nagel™ Supplier No. 740499.250S
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement columns for use with NucleoSpin™ Plasmid (NoLid) kits for rapid, small-scale preparation of high-purity, sequencing-grade plasmid DNA.

NucleoSpin™ Plasmid (NoLid)

  • For rapid small scale preparation high-purity plasmid DNA
  • Fast procedure – 6 Mini preps in 25 min
  • High capacity – up to 50 μg of plasmid DNA
  • Chaotropic salt wash buffer – extra pure DNA
  • Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
    • High-copy plasmids from E. coli
    • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
    • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
    • Supplementary protocol for M13 DNA availabl

Specifications

Color Clear
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 50 μg
Elution Volume 50 μL
Final Product Type Sequencing-grade plasmid DNA
For Use With (Equipment) NucleoSpin™ Plasmid (NoLid) Kit
For Use With (Application) Preparation of high-purity plasmid DNA
Includes Columns only
Product Type NucleoSpin Plasmid (NoLid) Column
Quantity 250 Pk.
Sample Volume 2 to 10 mL
Afficher plus Afficher moins
Identifiant produit
  • NucleoSpin Plasmid (NoLid)
Informations supplémentaires
  • MIXTURE LIST-Contain: sodium hydroxide solution (diluted < 2 %); guanidine hydrochloride; 2-propanol;Rnase
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.