missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid Transfection-grade, Mini kit for ultrapure plasmid DNA

Product Code. 15503636
Change view
Click to view available options
No. of Samples:
50
250
Unit Size:
250 reactions
50 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples Unit Size
15503636 50 50 reactions
15593626 250 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15503636 Supplier Macherey-Nagel™ Supplier No. 740490.50
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    107.10 € / 50 reactions
    Save 29.90 €
    21% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A fast way to purify transfection-grade plasmid DNA using mini-format technology to reduce endotoxins from bacterial lysates. Complete kit includes mini spin columns for an ultra-fast workflow.

Endotoxins are co-purified during typical plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. NucleoSpin™ Plasmid Transfection-grade kits are designed to reduce endotoxins from bacterial lysates.

  • Purifies high-copy plasmids for transfection
  • Novel technology to diminish endotoxin content (< 50 EU/μg DNA) for successful transfection
  • Rapid purification from E. coli via manual, vacuum or centrifugation processing
  • Ultra-fast procedure using mini spin columns simplifies the work-flow
  • Downstream applications: transfection of common cell lines (lipofection, electroporation)
  • Also suitable for standard molecular biology application, such as cloning and sequencing

Specifications

Bacterial Endotoxins <50 EU/ng DNA
Content And Storage Room temperature (18 - 25 °C)
Format Mini Spin Column
Isolation Technology Silica Membrane and Endotoxin-reduction Technology
Sample Type Bacterial Culture (E. coli, High-copy Plasmids)
Efficiency Binding capacity: 35 μg
Elution Volume Elution Volume: 30 - 50 μL
Final Product Type Transfection-grade plasmid DNA
For Use With (Equipment) Centrifuge, Vacuum manifold (e.g. NucleoVac 96)
For Use With (Application) Rapid purification of plasmids for transfection
Includes NucleoSpin Plasmid Transfection-grade Columns, Collection Tubes, Buffers, RNase A
No. of Samples 50
Purity A260/A280: 1.80 - 1.85
Sample Volume Up to 5 mL
Size < 15 kbp
Test Time 14 min/6 preps
Yield 15 - 30 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.