missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA, Mini kit for RNA purification

Product Code. 15220023
Change view
Click to view available options
No. of Reactions:
10
12
50
240
250
Format:
Mini Spin Column
QIAcube
Unit Size:
10 reactions
250 reactions
300mL
50 reactions
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Format unitSize
15220023 240 QIAcube Each
12373368 250 Mini Spin Column 250 reactions
11922402 50 Mini Spin Column 50 reactions
11912512 10 Mini Spin Column 10 reactions
15373604 12 QIAcube 300mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15220023 Supplier Macherey-Nagel™ Supplier No. 740955.240C
  • ONLINE EXCLUSIVE PRICE
    1465.00 €
    1443.75 € / Pack of 1
    Save 21.25 €
    1% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mini spin kit for isolation of RNA of highest integrity.

Features
  • DNase included for on-column DNA digestion– easy and efficient removal of contaminating DNA
  • NucleoSpin™ Filters (shredders) – efficient sample homogenization and reduction of viscosity
  • Up to 70 μg ready-to-use RNA
  • Parallel purification of genomic DNA by using the NucleoSpin™ RNA/DNA Buffer Set
Applications
  • RNA isolation from cultured cells, tissue (standard protocol)
  • RNA from < 109 bacterial cells (Gram-negative, Gram-positive) or < 108 yeast cells
  • RNA from ≤ 100 μL biological fluids
  • RNA clean-up from reaction mixtures
  • RNA from samples stored in RNAlater™
  • RNA from saliva samples collected with Oragene™ RNA (Genotek)
  • Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays

Specifications

Bacterial Content 70 μg
Detection Method Silica-membrane
Format QIAcube
Isolation Technology Silica Membrane
DNase Yes
Final Product Type NucleoSpin™ RNA Kit
For Use With (Application) RNA isolation from cultured cells, tissue
No. of Reactions 240
No. of Samples 4
Product Type NucleoSpin™ RNA Kit
Quantity 1 Each
Sample Size <5 x 106 cultured cells, <30 mg human/animal tissue, <109 bacterial cells, <108 yeast cells
Yield 14μg HeLa cells, 70μg Bacterial Cells
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate, ethanol (diluted < 20 %) / guanidine hydrochloride, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.