missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Plus XS, Micro kit for RNA purification with DNA removal column

Product Code. 15761639
Change view
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15761639 50 Preps 50 reactions
15741639 10 Preps 10 reactions
15751639 250 Preps 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15761639 Supplier Macherey-Nagel™ Supplier No. 740990.50
  • ONLINE EXCLUSIVE PRICE
    478.00 €
    407.40 € / 50 reactions
    Save 70.60 €
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for efficient and quick RNA isolation from the smallest samples of cells and tissues. State-of-the-art, dual-column kit includes gDNA removal columns and RNA binding columns.

NucleoSpin™ RNA Plus XS

  • Developed for extra small samples down to single cell analysis
  • Efficient and quick RNA isolation from cells and tissues
  • Two-column system includes gDNA removal columns and RNA binding columns
  • NucleoSpin™ gDNA Removal Columns – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal on one column in one step
  • New efficient lysis buffer – no b-mercaptoethanol or TCEP necessary
  • Variety of sample materials – cultured cells, animal tissue, plant tissue, microorganisms
  • Downstream applications include real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays

Specifications

Color Yellow/Blue ring
Content And Storage Room temperature (18-25 °C)
Format Mini Spin Column
Isolation Technology Silica Membrane
Elution Volume 5 - 30 μL
Sample Type Cultured Cells, Human or Animal Tissue
Efficiency Binding capacity: 110 μg
Final Product Type RNA (with typical RIN > 8)
For Use With (Equipment) Centrifuge
For Use With (Application) Isolation of RNA
Includes NucleoSpin RNA Plus XS Columns, NucleoSpin gDNA Removal Columns XS, Collection Tubes (2 mL), Collection Tubes (1.5 mL), Buffers
No. of Reactions 50 Preps
Product Type RNA Isolation Kit
Purity A260/A280: 1.9 - 2.2
Sample Size 1 - 105 cultured cells; < 5 mg human / animal tissue
Test Time 18 min/6 preps
Yield 0.05 to 0.2 ng (101 HeLa cells ), 0.5 to 2 μg (105 HeLa cells), 2.5 to 8 ng (0.5 μg mouse liver), 0.1 to 0.5 ng (0.5 μg mouse brain)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Plus XS
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate/guanidinium thiocyanate,ethanol (diluted < 20 %)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.