missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA XS, Micro kit for RNA purification

Product Code. 12723580
Change view
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12723580 50 50 reactions
12703580 10 10 reactions
12713580 250 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12723580 Supplier Macherey-Nagel™ Supplier No. 740902.50
  • ONLINE EXCLUSIVE PRICE
    481.00 €
    386.40 € / 50 reactions
    Save 94.60 €
    19% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purify high quality RNA quickly and easily. NucleoSpin™ RNA XS Columns enable purification of highly concentrated RNA from the smallest samples.

Features
  • Isolation of RNA from small sample quantities like biopsy material or single cells
  • Excellent RNA recovery and integrity
  • Concentrated RNA for sensitive downstream applications by elution in as little as 5 μL
  • rDNase included for on-column DNA removal
  • NucleoSpin™ Filters (shredders) – efficient homogenization and reduction of viscosity
  • High quality RNA, ready to use for RT-PCR and other applications
  • Reducing agent TCEP – no ß-mercaptoethanol necessary
Applications
  • RNA isolation from cultured cells
  • RNA isolation from tissue
  • RNA isolation from cryosections
  • RNA isolation from laser captured cells
  • RNA isolation from small amounts of plant material
  • RNA isolation from samples stored in RNAlater™
  • Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology,
  • RNase protection assays

Specifications

Format Mini Spin Column
Final Product Type NucleoSpin™ RNA XS Column
For Use With (Application) For RNA isolation from cultured cells, tissue, cryosections and laser captured cells
Includes Collection tubes, NucleoSpin™ filters, collection tubes (2mL), Collection tubes (1.5mL), buffers, RNase-free rDNase, carrier RNA, reducing agent TCEP
No. of Reactions 50
Product Type NucleoSpin™ RNA XS Column
Quantity 1 Each
Yield 0.1 to 1.5 ng (102 HeLa cells), 10 to 15 ng (103 HeLa cells), 100 to 150 ng (104 HeLa cells), 1000 to 1500 ng (105 HeLa cells)
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • NucleoSpin RNA XS
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Ergänzende Informationen
  • MIXTURE LIST-Contain: guanidinium thiocyanate, ethanol (diluted < 20 %) (denaturedwith 1%IPA/1%MEK, acc.2016/1867/EU); tr
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.