missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Tissue, Mini kit for DNA from cells and tissue

Product Code. 11982262
Change view
Click to view available options
No. of Reactions:
10
12 Preps
250
50
Unit Size:
10 reactions
12 preps
250 reactions
50 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11982262 250 250 reactions
11902512 10 10 reactions
15343604 12 Preps 12 preps
10498961 50 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) Chem class:3 Pack Group III
Product Code. 11982262 Supplier Macherey-Nagel™ Supplier No. 740952.250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) Chem class:3 Pack Group III

Mini spin kit for purification of DNA from a broad range of samples

  • Versatile kit for the isolation of genomic DNA from a wide range of different starting materials (cells, tissue, forensic and clinical samples)
  • 16 support protocols available in the user manual for highest flexibility
  • Highly sensitive membrane for reliable purification of even trace amounts of DNA
  • Rapid and easy protocol (20 min./prep excl. lysis)

Specifications

Cell Type Animal & Human tissue, cells
Capacity 60 μg
Column Type Mini spin column
Content And Storage Harzadous, 18°C to 25°C
Description Kit for purification of DNA from a broad range of samples
Isolation Technology Silica Membrane Technology
Sample Type Animal and Human Tissue, Cells
Elution Volume 60 to 100 μL
Final Product Type DNA
For Use With (Equipment) Centrifugation, Vacuum
For Use With (Application) Genotyping, Multiplex PCR, Next Generation Sequencing
Includes NucleoSpin Tissue Columns, Collection Tubes, buffers, Proteinase K
Key Functions DNA purification
No. of Reactions 250
Nucleic Acid Type DNA
Product Type DNA purification Kit
Quantity 1 Each
Purity A260/A280 1.7 - 1.9
Sample Volume <25 mg human/animal tissue, 102-107 cultured cells
Shipping Condition Room Temperature
Size 200 bp to 50 kbp
Source Animal & Human tissue, cells
Target DNA
Technique Silica-membrane technology
Test Time 20 min/prep (excl. lysis)
Yield 20 to 35μg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.