missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ totalRNA FFPE, Mini kit for RNA from FFPE

Product Code. 15297453
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15297453 1 Ea. 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15297453 Supplier Macherey-Nagel™ Supplier No. 740982.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mini spin kit for isolation of small and large RNA from formalin-fixed, paraffin-embedded samples.

Specifications

Format Mini Spin Column
Final Product Type NucleoSpin™ Total RNA FFPE Column
For Use With (Application) Rapid isolation of RNA from formalin-fixed, paraffin-embedded samples
Includes Collection Tubes (2mL), Collection Tubes (1.5mL), Paraffin Dissolver, buffers, Proteinase K, RNase-free rDNase
No. of Reactions 10
Product Type NucleoSpin™ Total RNA FFPE Column
Quantity 1 Ea.
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin totalRNA FFPE (10)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: rDNAse, paraffin disolver, 1,4-dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.