missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Bioanalysis™ Nucleospin TotalRNA Ffpe

Product Code. 15217463
Change view
Click to view available options
Volume (Metric):
10 sections (10 μm) with < 50 mg of tissue
Unit Size:
250 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15217463 10 sections (10 μm) with < 50 mg of tissue 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15217463 Supplier Macherey Nagel Bioanalysis™ Supplier No. 740982.250
  • ONLINE EXCLUSIVE PRICE
    1515.00 €
    1401.96 € / 250 reactions
    Save 113.04 €
    7% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mini spin kit for isolation of small and large RNA from formalin-fixed, paraffin-embedded samples

250 preparations, Macherey-Nagel

Specifications

Content And Storage 18°C to 25°C
Format Mini Prep
Sample Type FFPE, Fixed Samples, Tissue Sections
For Use With (Application) Isolation of Total RNA
No. of Reactions 250 Preps
Size > 200 nt
Target RNA
Volume (Metric) 10 sections (10 μm) with < 50 mg of tissue
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin totalRNA FFPE (250)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: rDNAse, paraffin disolver, 1,4-dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.