Learn More
Macherey-Nagel™ NucleoSpin™ Virus, Mini kit for viral RNA/DNA purification
Product Code. 13148496
Change view
Click to view available options
Quantity:
10 preps
50 preps
Unit Size:
10 reactions
50 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13148496 10 preps 10 reactions
13168496 50 preps 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13148496 Supplier Macherey-Nagel™ Supplier No. 740983.10
  • ONLINE EXCLUSIVE PRICE
    68.10 €
    60.48 € / 10 reactions
    Save 7.62 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Mini spin kit for time saving parallel isolation of viral RNA / DNA from different samples.
Specifications
Quantity 10 preps
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin Virus
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Supplemental information
  • MIXTURE LIST-Contain: proteinase K, liquid(origin: tritirachium album) / guanidine hydrochloride / 2-propanol