missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Virus, Mini kit for viral RNA/DNA purification

Product Code. 13168496
Change view
Click to view available options
Quantity:
10 preps
50 preps
Unit Size:
10 reactions
50 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13168496 50 preps 50 preps
13148496 10 preps 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13168496 Supplier Macherey-Nagel™ Supplier No. 740983.50
  • ONLINE EXCLUSIVE PRICE
    304.00 €
    263.55 € / 50 preps
    Save 40.45 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mini spin kit for time saving parallel isolation of viral RNA / DNA from different samples.

Specifications

Quantity 50 preps
Sample Volume <200 μL (up to 400 μL with two loading steps)
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin Virus
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Supplemental information
  • MIXTURE LIST-Contain: proteinase K, liquid(origin: tritirachium album) / guanidine hydrochloride / 2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.