Learn More
Hamilton™ Nueros™ 33 Gauge Replacement Needles
Product Code. 15655938
Change view
Click to view available options
Needle Gauge:
33 G
Unit Size:
Pack of 6
Product Code. Needle Gauge Unit Size
15655938 33 G Pack of 6
1 options
Product Code. 15655938 Supplier Hamilton™ Supplier No. 6546102
  • ONLINE EXCLUSIVE PRICE
    235.00 €
    222.27 € / Pack of 6
    Save 12.73 €
    5% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Autoclavable Autoclavable
Needle Gauge 33 G
Length (English) Needle 3.03 in.
Needle Point Style 4