Learn More
Syngene NuGenius9 XE
Product Code. 31749207 Shop All Syngene Products
Change view
Click to view available options
Pixel Size:
9 MP
Unit Size:
1 set
Product Code. Pixel Size Unit Size
31749207 9 MP 1 set
1 options
Product Code. 31749207 Supplier Syngene Supplier No. Nugenius9XE
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
NuGenius XE offers flexible DNA, stain free, autorads analysis. Intuitive touch screen system. Compact benchtop size with built in PC.
  • NuGenius XE offers flexible DNA, stain free, autorads analysis.
  • Intuitive touch screen system.
  • Compact benchtop size with built in PC.
  • Internal lighting options includes a UV transilluminator (purchased separately) for working with DNA gels (Ethidium bromide).
  • Our new UV-blue light converter screen (purchased separately) allows imaging of a wide range of safe dyes.
  • A visible converter option (purchased separately) can quickly extend its use for working with visible gels and blots (Coomassie blue and Silver stain).
  • The overhead LED white lighting is included as standard for easy sample positioning and focussing.
  • Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
  • With a 5MP camera and f/1.2 motor driven lens provides exquisite pixel resolution and unrivalled sensitivity for gels and blots with a viewing area of 20 x 24 cm
  • Images are saved directly to USB or network for easy transfer.
  • Quickly generate quantitative data with GeneTools analysis software.
  • NuGenius XE comes with unlimited copies of GeneTools.
Specifications
Pixel Size 9 MP
For Use With (Application) Gel Documentation
Product Type Gel Imaging System
Display Type Touchscreen