Learn More
Thermo Scientific™ Nunc™ 1.8 mL Externally-Threaded, Sterile Universal Tubes, Bulk pack
Product Code. 31935692
Change view
Click to view available options
Closure Color:
Blue
Green
Orange
Purple
Red
Yellow
Barcode:
2D
Non-coded
Packaging:
10 Bags of 48 Tubes
10 Racks of 48 Tubes
Unit Size:
Pack of 480
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Barcode Packaging unitSize
31935692 Blue Non-coded 10 Bags of 48 Tubes Pack of 480
31935661 Red 2D 10 Bags of 48 Tubes Pack of 480
31935548 Orange 2D 10 Bags of 48 Tubes Pack of 480
31935657 Yellow 2D 10 Bags of 48 Tubes Pack of 480
31935672 Green 2D 10 Bags of 48 Tubes Pack of 480
31935563 Blue 2D 10 Bags of 48 Tubes Pack of 480
31935614 Purple 2D 10 Bags of 48 Tubes Pack of 480
31935574 Red Non-coded 10 Bags of 48 Tubes Pack of 480
31935629 Orange Non-coded 10 Bags of 48 Tubes Pack of 480
31935566 Yellow Non-coded 10 Bags of 48 Tubes Pack of 480
31935560 Green Non-coded 10 Bags of 48 Tubes Pack of 480
31935544 Purple Non-coded 10 Racks of 48 Tubes Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 31935692 Supplier Thermo Scientific™ Supplier No. 374503BLU
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
Use the automation friendly tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available in clear and color-coded caps, racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.
Use the automation friendly Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes to improve the efficiency and throughput of your sample storage workflow. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.
Scalable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10 x 10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
  • 374502NOV is featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Autoclavable Not Autoclavable
Bottom Shape Flat
Material Polypropylene
Thread Style External
Packaging 10 Bags of 48 Tubes
Barcode Non-coded
Sterility Sterile
Graduated Yes
Product Type Cryogenic Storage Tube