Learn More
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps.
138.00 € - 214.66 €
Product Code | Brand | Culture Area | Neck Style | Barcode | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Neck Style | Barcode | Price | Quantity & Availability | |||
10205232
|
Thermo Scientific™
178983 |
175 cm2 | Straight | Large Code 128 |
214.66 €
Pack of 32 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
12344183
|
Thermo Scientific™
178985 |
175 cm2 | Straight | Yes |
174.00 €
Pack of 32 |
In Stock
Log in to see stock. |
|||
12304193
|
Thermo Scientific™
178885 |
175 cm2 | Straight | No |
138.00 €
Pack of 32 |
In Stock
Log in to see stock. |
|||
10127340
|
Thermo Scientific™
178883 |
175 cm2 | Straight | No |
165.00 €
Pack of 32 |
In Stock
Log in to see stock. |
|||
10170961
|
Thermo Scientific™
178905 |
80 cm2 | Straight | No |
146.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. The filter caps allow constant airflow and minimize chances of contamination.
Features:- Nunclon Delta Certification including testing on four different cell types is provided in each case.
- Short wide necks, straight side walls and 1/3-turn caps are optimized for robotic operation.
- Filter caps ensure constant gas exchange.
- Barcoded 175cm2 flasks with large Code 128 are compatible with automation.
- Sterilized by gamma irradiation and certified to be non-pyrogenic.
Specifications
Sterile | |
Filter |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.