Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System

Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
- Ideal for adherent cells
- Boost production efficiency with more surface area in a small footprint
- Reduce contamination risks with a closed system
Specifications
Specifications
| Description | Original Cell Factory |
| Material | Polystyrene |
| No. per Case | 8/Cs. |
| No. per Pack | 1 Ea. |
| Product Type | Cell Factory System |
| Surface Coating | Nunclon™ Delta |
| Green Features | ISCC PLUS |
| Closure Type | Adapter Cap and Port Cover Cap |
| Culture Area | 632 cm² |
| Surface Treatment | Nunclon Delta |
| Show More |
Frequently Asked Questions (FAQs)
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .
We recommend a 200 mL volume of medium per tray (632 cm^2).
Therefore the minimum volumes we recommend are as follows:
Cat. No.; No. per Pack/No. per Case; No. of Trays; Minimum Volume (mL)
- 165250; 1/8; 1-Tray; 200 mL
- 167695; 1/5; 2-Tray; 400 mL
- 164327; 1/2; 10-Tray; 2000 mL
- 170009; 1/6; 10-Tray; 2000 mL
- 139446; 1/2; 40-Tray; 8000 mL
Please review the following information regarding the dimension and culture areas for each tray version of the Nunc Cell Factory Systems:
Cat. No.; No. per Pack/No. per Case; No. of Tray Layers; Dimensions L x W x H (mm); Culture Area (cm^2)
- 165250; 1/8; 1-Tray; 335 mm x 205 mm x 37 mm; 632 cm^2
- 167695; 1/6; 2-Tray; 335 mm x 205 mm x 52 mm; 1264 cm^2
- 164327; 1/2; 10-Tray; 335 mm x 205 mm x 190 mm; 6320 cm^2
- 170009; 1/6; 10-Tray; 335 mm x 205 mm x 190 mm; 6320 cm^2
- 139446; 1/2; 40-Tray; 335 mm x 205 mm x 700 mm; 25284 cm^2
Due to the height of the 10-tray and 40-tray Nunc Cell Factory Systems, cells cannot be viewed with common microscopes. Typically cells are seeded in a 1-tray or 2-tray cell factory system as a control. Cells grown in each of these products can be viewed under a microscope.
Alternatively, inverted stereo microscopes, with a powerful light source on the view side, and adjusted for the height of the cell factory system have been used to view the first few layers.
Our Nunc Cell Factory Systems are designed for large scale cell culture and production of biomaterials such as vaccines, monoclonal antibodies, etc. They provide large growth surface in limited space areas and have the Nunclon Delta surface coating. Some benefits are the low risk of contamination and the easy to handle multi-layer stacks. Nunc Cell Factory Systems are available with 1-, 2-, 10-, and 40-tray versions.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.