Learn More
Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface
Descrizione
Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
Specifica
Specifica
| Autoclavable | Not Autoclavable |
| Bottom Shape | Flat |
| Color | Clear |
| Culture Area | 145 cm2 |
| Disposable | Single-use |
| Gridded | Not Gridded |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Cell Culture Dish |
| Vedi altri risultati |
For Laboratory Use
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.