missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface

Product Code. 30408779
Change view
Click to view available options
Culture Area:
175 cm²
25 cm²
75 cm²
Closure Type:
Filter
Solid
Unit Size:
Pack of 100
Pack of 20
Pack of 200
Pack of 30
Pack of 6
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Closure Type Unit Size
30408779 75 cm² Solid Pack of 100
30408805 25 cm² Filter Pack of 200
17987432 25 cm² Filter Pack of 20
30408801 25 cm² Solid Pack of 200
17997432 25 cm² Solid Pack of 20
30408795 75 cm² Filter Pack of 100
17917432 75 cm² Solid Pack of 20
17937432 175 cm² Filter Pack of 6
30408785 175 cm² Filter Pack of 30
30408806 175 cm² Solid Pack of 30
17977432 175 cm² Solid Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 30408779 Supplier Thermo Scientific™ Supplier No. 156378
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY

Specifications

Description Nunc EasYFlask, 75 cm²
For Use With (Application) General cell culture, primary cell expansion, cell therapy research
Neck Style Angled
No. per Case 100 Flasks
No. per Pack 5 Flasks
Product Type Cell Culture Flask
Mouth Style Wide
Sterility Sterile
Closure Type Solid
Culture Area 75 cm²
Surface Treatment Nunclon Supra
Quantity 100/Cs.
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.