Learn More
Thermo Scientific™ Nunc™ IVF ICSI Dish
Product Code. 11327704
Change view
Click to view available options
Diameter (Metric):
51 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
11327704 51 mm Each
1 options
Product Code. 11327704 Supplier Thermo Scientific™ Supplier No. 150265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc™ IVF ICSI Dishes enable clear viewing of samples with a super-polished optical center and meet the exacting requirements of IVF testing, certification, and validation.
Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc IVF ICSI Dishes meet the exacting requirements of IVF testing, certification, and validation.
Innovative features for quality and ease-of-use
  • Maintain optimal temperature with true flat bottom
  • Low profile provides optimal angles during ICSI process
  • Clearly see sample with super polished optical center
  • Increased safety with easy grip design
  • Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF
  • CE-marked (Notified Body 2797), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Certificates of conformity for your assurance on every lot
Sterile (SAL 10-6)
  • Non-pyrogenic (LAL test)
  • USP Class VI test
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE-marked, FDA-cleared, MEA-tested
Description IVF ICSI Dish, Lid w/ Airvent
Bottom Shape Flat
Color Clear
Material Polystyrene (USP Class VI)
Sterility Sterile
Surface Treatment Non-treated
Type Dish
Well Count 1
No. per Case 120
No. per Pack 1
Height (Metric) 9 mm
Diameter (Metric) 51 mm
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.