missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ IVF Dishes
BIOPHARMA

Product Code. 10262623
Change view
Click to view available options
Diameter (Metric):
35 mm
54 mm
88 mm
Height (Metric):
11 mm
14 mm
15 mm
Culture Area:
21.5 cm2
56.7 cm2
8.8 cm2
Unit Size:
Pack of 150
Pack of 400
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) Culture Area Unit Size
10262623 54 mm 14 mm 21.5 cm2 Pack of 400
10170753 35 mm 11 mm 8.8 cm2 Pack of 500
10021963 88 mm 15 mm 56.7 cm2 Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10262623 Supplier Thermo Scientific™ Supplier No. 150270
  • ONLINE EXCLUSIVE PRICE
    344.00 €
    276.47 € / Pack of 400
    Save 67.53 €
    19% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.

Carry out IVF applications using high-quality, reliable Thermo Scientific™ Nunc™ IVF Dishes.

Product features for reliable results:

  • Facilitate gas exchange with vented lid
  • Available in 35mm, 60mm and 90mm sizes
  • Certificates of conformity available for every lot

Rigorously tested, certified and validated for IVF:

  • CE-marked (Notified Body 2460), FDA-listed
  • Produced under ISO 13485 with full lot traceability
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL tested)
  • USP Class VI polystyrene
  • Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested
Description IVF Dish, 60mm, Lid w/Airvent
Culture Area 21.5 cm2
Material Polystyrene (USP Class VI)
Sterility Sterile
Surface Treatment Untreated
No. per Case 400
No. per Pack 10
Height (Metric) 14 mm
Diameter (Metric) 54 mm

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.