missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Multidishes with UpCell™ Surface

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.

Brand:  Thermo Scientific™ 174900

172.24 EUR valid until 2024-12-31
Use promo code "21928" to get your promotional price.



Code : 76

Additional Details : Weight : 0.46800kg

 View more versions of this product

Product Code. 10288143

  • 255.00 € / Pack of 6
In Stock
to see stock.
Add to basket

Description

Description

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of "cell sheet" with the help of a supportive membrane.

Traditional cell harvesting by enzymatic and mechanical methods often compromises the integrity of surface proteins and the viability of harvested cells. By contrast, UpCell Surface allows cell harvesting by simply reducing the temperature of the cell culture to below 32°C without the need of enzyme, resulting in highly viable cells with intact cell surface proteins.
  • No trypsinization—preserve cell surface proteins
  • No physical force—get high cell viability
  • For culture passaging, single-cell analyses and cell transplantation research
  • 6-well multidish comes with supportive membranes that enable harvesting of cell sheets and creations of 3-D tissue models
  • Minimal hands-on time
  • Customized bar coding is available

Recommended for:

Culture passaging; Single-cell analyses, Cell transplantation research, 3D tissue models

Specifications

Specifications

12 well, UpCell
2 mL (Well)
Multi Dish
6
UpCell, Temperature Responsive
12
3.5 sq. cm
2 mL
1
Clear
Sterile
Rectangular, Flat Bottom (F-well)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only