missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ PETG Roller Bottles

Product Code. 10385073
Change view
Click to view available options
Cell Growth Area:
1050 cm²
1700 cm²
1800 cm²
2100 cm²
4200 cm²
Bottle Surface:
Pleated
Smooth
Packaging:
20/Cs.
22/Cs.
5/Sleeve, 20/Cs.
Unit Size:
Pack of 20
Pack of 22
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cell Growth Area Bottle Surface Packaging Unit Size
10385073 1800 cm² Smooth 22/Cs. Pack of 22
10059702 1050 cm² Smooth 5/Sleeve, 20/Cs. Pack of 20
10712974 1050 cm² Smooth 20/Cs. Pack of 20
10263393 1050 cm² Smooth 5/Sleeve, 20/Cs. Pack of 20
10627584 1700 cm² Pleated 20/Cs. Pack of 20
10704144 2100 cm² Pleated 20/Cs. Pack of 20
10551844 2100 cm² Pleated 5/Sleeve, 20/Cs. Pack of 20
10596895 4200 cm² Pleated 22/Cs. Pack of 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10385073 Supplier Thermo Scientific™ Supplier No. 186022
  • ONLINE EXCLUSIVE PRICE
    535.00 €
    366.00 € / Pack of 22
    Save 169.00 €
    31% Off
View all promotions
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.

Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.

Key Features:

  • Molded of durable PETG
  • Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
  • Provides solutions for all scales with wide range of surface areas from 1050 cm² to 4200 cm²
  • Reduces wrist strain and increases productivity with quick-action ergonomic closure
  • Can be traced by lot number on every bottle

Recommended for:

  • Cell culture expansion
  • Vaccine production
  • Antibody production

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Bottle Surface Smooth
Capacity (Metric) 5 L
Color Clear
Description 1860-22
Material PETG
No. per Case 22/Cs.
No. per Pack 22 Pk.
Packaging 22/Cs.
Sterility Sterile
Cell Growth Area 1800 cm²
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Type Quick-Action Ergonomic
Capacity (English) 67.63 fl. oz.
Shape Round
Disposable Single-use
Product Type Roller Bottle
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.