missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm

Product Code. 10175162
Change view
Click to view available options
Wells:
12 + 2-well
12 + 2-well, w/adapters
20-well
20-well, w/adapters
26-well
26-well, w/adapters
Unit Size:
1 set
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Unit Size
10175162 12 + 2-well 1 set
10378242 12 + 2-well, w/adapters 1 set
10175212 20-well Pack of 10
10388242 20-well, w/adapters 1 set
10115312 26-well 1 set
10277042 26-well, w/adapters 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10175162 Supplier Invitrogen™ Supplier No. WG1201BOX
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

One box of 10 gels

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Features of NuPAGE Bis-Tris gels:
• Better protein integrity—optimized sample preparation process preserves your proteins
• Wide ranges of molecular weight separation—select the right gel and running buffer to get the optimal separation of your proteins
• Faster run times—get separation of your proteins in as little as 35 minutes
• Longer shelf life—NuPAGE Bis-Tris gels can be stored for at least 12 months at room temperature

Choose the right NuPAGE Bis-Tris gel for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

NuPAGE Bis-Tris Midi gels can be used with the XCell SureLock Midi Cell gel apparatuses (WR0100) or conveniently with the Bio-Rad Criterion Cell using our adapters (WA0999).

For transfer of proteins to a membrane, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001). Alternatively, traditional wet transfer can be performed using the Bio-Rad Criterion Cell using NuPAGE Transfer buffer (NP00061).

TRUSTED_SUSTAINABILITY

Specifications

Adapters No Adapters
Gel Percentage 10%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Recommended Applications Denaturing
Sample Loading Volume Up to 45 μL + 15 μL
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 12 + 2-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Room Temperature
Storage Requirements Store at 4–25°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 13 cm
Product Line NuPAGE
For Use With (Equipment) XCell4 SureLock Midi-Cell
Show More Show Less
Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Do I need to add water or buffer to the Displacement Dam when I run one or two gels in the XCell4 SureLock Midi-Cell?

The XCell4 Displacement Dam is used when fewer than 3 gels are run in the XCell4 SureLock Midi-Cell. The Displacement Dam is placed in the lower buffer chamber instead of a second buffer core. Do not add any buffer or water to the Displacement Dam. However, if water or buffer is accidentally added to the Displacement Dam, it does not affect the electrophoresis run.

What are the maximum load volumes for the NuPAGE Midi Gels?

Recommended maximum load volumes:
12 + 2-well gel: 45 µL in sample well, 15 µL in marker well
20-well gel: 25 µL
26-well gel: 15 µL

What do I need to run the NuPAGE Invitrogen Midi Gels in the Bio-Rad Criterion Cell?

You will need a Midi Gel Adapter. The Midi Gel Adapter is a self-adhesive, upper buffer chamber used when running the NuPAGE Invitrogen Midi-Gel in a Bio-Rad Criterion Cell. The Midi Gel Adapter may be filled with up to 60 mL of buffer. The Midi Gel Adapters are available packaged with the Midi-Gels (Invitrogen and NuPAGE Invitrogen Midi Gels with Cat. Nos. that end in "A") and also on their own in packages of 10 (Cat. No. WA0999). Running the NuPAGE Midi-Gels in the XCell4 SureLock Gel Apparatus does not require the use of the Midi Gel Adapter.

Are the NuPAGE Midi Gels compatible with multichannel pipettes?

Multichannel pipette compatibility: 12+2-well (8- and 12-channel pipette-compatible) and 26-well (8- and 12-channel pipette-compatible).
The 12-channel pipettes work with the 26-well gels. You will need to alternate lanes (e.g., aligns with every two lanes). There will be two extra lanes (for MW markers). The 12-channel pipettes work with the 12+2 well gels as well.

How much of the pAd/CMV/V5-DEST or pAd/PL-DESTpAD destination vector do I need to add in the LR reaction?

One should use 2 µl = 300 ng pAd-DEST vector. Please Note that Version D of the pAd/CMV/V5-DEST and pAd/PL-DEST Gateway Vectors product manual (manual number 25-0544) had an error on page 9 regarding this protocol. The manual instructed users to add 1 µl, but this has since been corrected to reflect the correct protocol of addition of 2 µl.

What is the advantage of NuPAGE Gels over regular Tris-Glycine gels?

The neutral operating pH of the NuPAGE Gels and buffers provides following advantages over the Laemmli system:
-Longer shelf life of 8-12 months due to improved gel stability
-Improved protein stability during electrophoresis at neutral pH resulting in sharper band resolution and accurate results (Moos et al, 1998)
-Complete reduction of disulfides under mild heating conditions (70 degrees C for 10 min) and absence of cleavage of asp-pro bonds using the NuPAGE LDS Sample buffer (pH > 7.0 at 70 degrees C)
-Reduced state of the proteins maintained during electrophoresis and blotting of the proteins by the NuPAGE Antioxidant
Please refer to the following paper: Moos M Jr, Nguyen NY, Liu TY (1988) Reproducible High Yield Sequencing of Proteins Electrophoretically Separated and Transferred to an Inert Support. J Biol Chem 263:6005-6008.

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

Can I use CTAB rather than SDS in my sample buffer?

No, CTAB will not work with any of our gels except for the NuPAGE Tris-Acetate gels. To use CTAB, you would need to use a running buffer of 50 mM acetic acid and 50 mM beta-alanine in equal concentrations. You would also need to switch the electrodes. Since CTAB is a cationic detergent, this would establish conditions for running a basic protein towards the anode (into the gel).

I ran my protein under native conditions on a Tris-Glycine gel. It has a pI that is higher than the pH of the Tris-Glycine transfer buffer. Can you offer some tips for transferring it?

- Increase the pH of Tris-Glycine transfer buffer to 9.2, allowing all the proteins below pI 9.2 to transfer towards the anode electrode.
- Use the Tris-Glycine transfer buffer and place a membrane on both sides of the gel. If there are any proteins that are more basic than the pH of the transfer buffer, they will be captured on the extra membrane placed on the cathode side of the gel. Both membranes can then be developed in the same manner.
- Prior to blotting, incubate the gel for 15 minutes in Tris-Glycine transfer buffer containing 0.1% SDS. The small amount of SDS will give the proteins enough charge to move unidirectionally towards the anode and in most cases, should not denature the protein. Proceed with the transfer using regular Tris-Glycine transfer buffer.

My gel run is faster than normal with poor resolution. What could be causing this problem?

Here are possible causes and solutions:

- Buffers are too concentrated or incorrect. Check buffer recipe; dilute or re-make if necessary.
- Voltage, current or wattage is set at a higher limit. Decrease power conditions to recommended running conditions.

My gel will not run. Can you please help me troubleshoot?

It is important to determine whether the problem is with the power supply, the apparatus or the gel. Often, it helps to switch out the power supply or the lid to see if there is a faulty contact. Also, check to see whether the tape from the bottom of the gel cassette has been removed and whether the buffer core is damaged. Additionally, make sure there is sufficient buffer in the electrophoresis tank to cover the wells of the gel.

Which of your protein gels can I run using the XCell4 SureLock Midi-Cell?

Our Midi gels can be run using the XCell4 SureLock Midi-Cell.

After western detection, my membrane has a lot of spots. What could have gone wrong?

Here are possible causes and solutions:

- Membrane blotting pads are dirty or contaminated. Soak pads with detergent and rinse thoroughly with purified water before use. Replace pads when they become worn or discolored.
- Blocking was uneven. The incubation dish must be sufficiently big to allow thorough coverage of membrane. Shake or agitate during each step.

I am getting a lot of non-specific binding after western detection. Can you offer some tips?

Here are possible causes and solutions:

- Membrane contaminated by fingerprints or keratin proteins: Wear clean gloves at all times and use forceps when handling membranes. Always handle membranes around the edges.
- Concentrated secondary antibody used: Make sure the secondary antibody is diluted as recommended. If the background remains high, but with strong band intensity, decrease the concentration of the secondary antibody.
- Concentrated Primary antibody used: Decrease the concentration of the primary antibody.
- Affinity of the primary antibody for the protein standards: Check with the protein standard manufacturer for homologies with primary antibody.
- Insufficient removal of SDS or weakly bound proteins from membrane after blotting: Follow instructions for membrane preparation before immunodetection.
- Short blocking time or long washing time: Make sure that each step is performed for the specified amount of time.

I am getting very high background after western detection. Can you please offer some tips?

Here are possible causes and solutions:

- Insufficient blocking or non-specific binding: We suggest trying our WesternBreeze Blocker/Diluent (Cat. No. WB7050).
- Membrane is contaminated: Use only clean, new membranes. Wear clean gloves at all times and use forceps when handling membranes.
- Higher intrinsic background with PVDF membranes: Switch to nitrocellulose membranes.
- Nitrocellulose membrane not completely wetted: Follow instructions for pre-wetting the membrane.
- Blot is overdeveloped: Follow recommended developing time and remove blot from substrate when signal - to -noise ratio is acceptable.
- Insufficient washing ; Follow recommended number of washes. In some cases, it may be necessary to increase the number or duration of washes.
- Concentrated secondary antibody used: Determine optimal antibody concentration by performing a dot blot and dilute antibody as necessary.
- Concentrated primary antibody used: Determine optimal antibody concentration by performing a dot blot and dilute antibody as necessary.

I am unable to visualize my protein bands after western detection. What is the problem?

Here are possible causes and solutions:

- The primary antibody and secondary antibody are not compatible: Use a secondary antibody that was raised against the species in which the primary antibody was raised.
- The primary antibody is too dilute: 1) Use a more concentrated antibody solution. 2) Incubate longer (e.g., overnight) at 4 degrees C. 3) Use fresh antibody and keep in mind that each time an antibody solution is used, its effective antibody concentration decreases.
- Something in your blocking buffer interferes with binding of the primary and/or secondary antibody: Try an alternate blocking buffer ± a mild surfactant like Tween-20 (0.01-0.05% v/v). There are many blocking buffer recipes available, based on non-fat dry milk, BSA, normal serum, gelatin and mixtures of these and other materials. Note that BSA (1-5%) is considered the best blocker for nitrocellulose membranes. It is easy to check the efficacy of different blocking buffers by performing dot-blots.
- The primary antibody does not recognize the protein in the species being tested: 1) Evaluate primary antibodies by dot-blotting first to how well they react with your protein. 2) Check the immunogen sequence, if provided, and determine if it is found in your protein. 3) If no immunogen sequence is available, perform a PubMed/BLAST alignment to assess the degree of homology between your target protein and the protein against which the antibody was generated. Note that many antibodies against human proteins will also recognize the non-human primate version because there is usually a high degree of amino acid identity. In contrast, many antibodies against human proteins will not recognize the corresponding proteins from rodents (and vice versa). Remember that significant homology between sequences does not guarantee that the antibody will recognize your protein. 4) Always run the recommended positive control, if available.
- Insufficient protein is bound to the membrane or the protein of interest is not abundant enough in the sample: 1) Load at least 20-30 ?g protein per lane on your gels (as a starting point), since proteins representing less than ~0.2% of the total protein are difficult to detect on western blots. 2) Use an enrichment step to increase the concentration of the target protein. For example, prepare two nuclear lysates prior to blotting nuclear proteins or perform an immunoprecipitation (IP) prior to SDS-PAGE. 3) Reduce the volume of cell extraction buffer used to lyse your cells or tissue. 4) Be sure to use freshly prepared protease inhibitors and phosphatase inhibitors, if needed, in your protein extraction buffer. 5) Run the recommended positive control, if available.
- Poor or no transfer of the proteins to the membrane 1) Check the protein transfer efficiency with a reversible protein stain like Invitrogen Reversible Membrane Protein Stain, ponceau S, amido black or use pre-stained molecular weight standards. 2) Verify that the transfer was performed with the correct electrical polarity. 3) Remember that proteins with basic pI values (e.g., histones) and high MW may not transfer well. 4) Remember that if your target protein has a low MW (≤10 kDa), it may transfer more quickly than expected. 5) If you are using PVDF membranes, make sure to pre-soak the membrane in methanol first before soaking it in transfer buffer. Note that methanol in transfer buffer increases protein binding to nitrocellulose, but omitting methanol can increase transfer efficiency of high MW proteins. 6) Low MW proteins may pass through the 0.45 µm pores in nitrocellulose membranes, so switch to NC with 0.2 or 0.1 µm pores instead.
- Excessive washing or blocking of the membrane:- 1) Avoid over-washing the membrane. Extra washing will not allow you to visualize your protein of interest if there are other problems with your blot. 2) Avoid over-blocking by using high concentrations of the blocking buffer components or long incubation times. Too much blocking can prevent your antibodies from binding to your protein. Gelatin, in particular, can mask proteins on the blot, so avoid it, if possible. Milk can also mask proteins, so instead of using 5% milk in your blocking buffer, try using it at 0.5% instead, or remove it altogether. 3) Switch to a different blocking reagent and/or block the blot for less time.
- Using the same solution of diluted primary antibody repeatedly: Use freshly-diluted antibody for each western blot because the effective concentration of a diluted antibody decreases each time it is re-used. Also, remember that dilute solutions of antibodies are less stable and may lose their activity rapidly.
- The enzyme conjugated to your secondary antibody is not working: 1) Make a fresh dilution of your secondary antibody conjugate each time you need it. Enzymes (and antibodies) may lose activity quickly in dilute solutions. 2) Omit sodium azide in buffers if you are using HRP-conjugated antibodies. 3) Avoid high heme concentrations (from blood contamination), which can interfere with HRP-based detection. 4) Avoid using phosphate in buffers with alkaline phosphatase-antibody conjugates because phosphate inhibits enzyme activity.
- Your colorimetric or other detection reagent is old and inactive: 1) Use fresh enzyme substrate for each experiment. 2) Don't use ready-to-use substrate reagents if they have changed color on their own or if they have passed their expiration date. 3) Do not dilute substrate solutions unless instructed to do so in the product manual.

I ran my protein sample on one of your gels and the bands look non-distinct and smeary after western detection. What should I do?

Here are some suggestions:

- Make sure that the correct amount of protein is loaded per lane; loading too much protein can cause smearing.
- Bands will not be as well resolved in low percentage gels; try using a higher percentage gel.
- This may be due to the antibody being too concentrated. We recommend following the manufacturer's recommended dilution or determining the optimal antibody concentration

I ran my protein samples on one of your protein gels and saw V-shaped protein bands. Can you please help me troubleshoot?

V-shaped protein bands are caused by the presence of DNA in the sample. The artifact might be eliminated or minimized by shearing the DNA with additional sonication after the SDS-solubilization step. Alternatively, the DNA can be removed from the sample using an ultra-centrifuge.

I have set up my NuPAGE gel to run and have switched on the power supply, but my gel is not running and there is no voltage or current reading on the power supply. What is wrong?

*Double check that the tape on the bottom of the gel has been removed.
*Make sure that the gel(s) are oriented so that the taller sides of the cassette (with the printing) are facing the outside of the electrophoresis unit.
*Make sure that the inner buffer chamber is filled sufficiently so that the wells are covered with buffer. If the wells are not covered, check for leaks and reseal.
*Double check to see if there are any loose electrodes or connections on the Mini cell unit.
*Check the power supply unit.

I am running my NuPAGE gel using the XCell SureLock Mini Cell but the leads do not fit into my power supply. Can you please help?

You may purchase the ZOOM adapters, Cat. No. ZA10001 to help you connect your leads to the power supply.

I am trying to load my samples on a NuPAGE gel but am not able to see the sample wells. Can you please suggest some tips?

We recommend marking the cassette at the bottom of the wells with a marker pen prior to assembling the upper buffer chamber. Also, we recommend illuminating the bench area with a light source placed directly behind the XCell SureLock unit.

I am running my NuPAGE gel using the XCell SureLock Mini Cell, and the gel run is taking longer than usual. What could be causing this?

Here are possible causes and solutions:

1) Buffers are too dilute: Check buffer recipe; remake if necessary.
2) Upper buffer chamber is leaking: Make sure the buffer core is firmly seated, the gaskets are in place and the gel tension lever is locked.
3) Voltage is set too low: Set correct voltage.

I had problems transferring my larger molecular weight proteins from my NuPAGE gel. Can you please offer some suggestions?

For proteins larger than 100 kDa, we recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing methanol and 0.01% SDS.

My samples are in RIPA buffer? Can I run them on a NuPAGE Bis-Tris gel?

RIPA buffer contains a lot of detergent and hence would not be compatible with NuPAGE gels.

I accidentally froze my NuPAGE Bis-Tris gels. Is it still okay to use them?

We do not recommend using frozen NuPAGE gels as:

*They may have become brittle and may crack either before or after the run, or even when in the staining solution.
*May cause the formation of bubbles during the run and band distortion particularly in the center lanes.

I ran my antibody sample on a NuPAGE Bis-Tris gel and saw a forward smear in all the lanes. What could have caused this?

A forward smear indicates that the antibody was being reduced in the gel during migration. This could have been caused by the addition of antioxidant in the sample buffer or due to rearrangement of disulfide bonds during heating in NuPAGE Sample buffer. Make sure that the correct concentration of reducing agent is used in the sample buffer and do not add any antioxidant in the sample buffer.

I ran my protein samples on a NuPAGE Bis-Tris gel and I see a bubble in the center of the gel with significant band distortion particularly in the center lanes. What is the reason?

This can happen if a frozen NuPAGE gel was used. Please make sure that NuPAGE gels are stored at 4-25 degrees C and are not accidentally frozen. Check refrigerator settings and store the gels on lower shelves away from the freezer section and away from the condenser.

The protein bands in some of my gel lanes are irregular or wavy? What would have caused this problem?

This could be due to:

*Debris in the well
*High salt in the sample (make sure that the salt concentration does not exceed 50-100 mM)
*Running buffer issue
*Gel casting error

I am seeing a very wavy and uneven dye front with my samples. Can you please help me troubleshoot?

This could be due to a gel polymerization issue combined with incorrect sample preparation (final sample dilution less than 1X). Please try a different lot of the same gel and make sure that the sample is correctly prepared.

I am seeing a faint, artifact doublet band at ~60 kDa in all my lanes. This band seems to be getting darker the longer I stain the gel. What could be causing this?

Possible cause:

*Excess reducing agent (beta-mercaptoethanol)
*Skin protein contaminants (keratin)

Remedy:

*The addition of iodoacetamide to the equilibration buffer just before applying the sample to the gel has been shown to eliminate these artifact bands.
*Use new electrophoretic solutions and wear gloves when handling and loading the gel. This issue is more common when highly sensitive stains are used.

I loaded different protein samples in each well but I see the same protein band in several neighboring lanes. What could have happened?

Possible cause:

*Carry-over contamination of sample from one well into neighboring wells due to loading error
*Contaminated running buffer
*Gel casting error: malformed wells

Remedy:

*Use a gel loading tip to load wells
*Reduce the sample volume
*Do not delay while loading wells
*Do not delay after the run, as proteins can diffuse horizontally; a full well left next to an empty well would eventually contaminate the empty well over time.

My protein bands appear to be skewed or distorted. What is the problem?

Possible cause:

*Poor polymerization around sample wells
*High salt concentration in sample
*Uneven gel interface
*Excessive pressure applied to the gel plates when the gel is placed into the clamp assembly
*Uneven heating of the gel
*Insoluble material in the gel or inconsistent pore size throughout the gel
*Air bubble during the run

Remedy:

*Remove excess salt/other material by dialysis, Sephadex G-25 or any other desalting column or using an Amicon concentrator.
*Either use a cooled apparatus or reduce the current at which electrophoresis is performed.

I ran my reduced protein samples under denaturing conditions and am seeing doublet protein bands when I expect to see single bands. Why is this happening?

A portion of the protein sample may have re-oxidized during the run, or may not have been fully reduced prior to the run. We recommend preparing fresh sample solution using fresh beta-mercaptoethanol or dithiothreitol (DTT). For NuPAGE gels, we recommend adding antioxidant to the running buffer.

My gel seems to be lifting off the cassette. What could be causing this?

Gel lifting off the cassette can be caused by:

*Expired gels that are degrading
*Improper storage of gels
*Too much heat accumulating during the electrophoresis run due to excessive current
*Insufficient polymerization of the polyacrylamide

I am seeing a faint shadow, or "ghost" band below a normal and expected protein band? What could be the potential issue?

Ghost bands are usually attributed to a slight lifting of the gel from the cassette, which results in the trickling down of some sample beyond its normal migration point. It then accumulates and appears as a faint second band.

My protein bands in the outer lanes of the gel show a "smiling" effect. Can you please help me troubleshoot?

"Smiling" bands may be the result of the acrylamide in the gel breaking down, leaving less of a matrix for the proteins to migrate. We recommend checking to ensure that the gels have not been used past their expiration date.

I see dumbbell or barbell shaped bands after protein electrophoresis. What could be causing this?

Barbell shaped bands are a result of loading too large of a sample volume. When a large sample volume is loaded, part of the sample tends to diffuse to the sides of the wells. When the run begins and the sample moves through the stacking portion of the gel, the sample will incompletely stack causing a slight retardation of the portion of the sample that diffused to the sides of the wells. This effect may be intensified for larger proteins, whose migration is more impeded in the low concentration acrylamide of the stacking gel. To alleviate the problem, we recommend concentrating the protein and loading a smaller volume. This gives a "thinner" starting zone.

Why do I get streaking forward or "frowning" of one of my samples on my protein gel?

Here are possible causes and solutions:

1) Sample overload: Do not overload samples
2) Addition of reducing agent that is not fresh: Reduce samples right before loading and do not use samples that have been stored in reducing agent
3)Re-oxidation of the protein during the run: Add antioxidant to the running buffer if you are running NuPAGE gels
4) Presence of highly hydrophobic regions where the protein can exclude SDS: Load the sample with 2X sample buffer instead of 1X sample buffer
5) Excess salt in the sample: Precipitate and reconstitute in lower salt buffer
6) Not enough SDS in the sample: Add SDS to the upper buffer chamber (try 0.1%, 0.2%, 0.3% and 0.4% SDS)

Can I use beta-mercaptoethanol instead of the NuPAGE Sample Reducing agent?

Although we recommend using the NuPAGE Sample Reducing agent for stability reasons, fresh, neat beta-mercaptoethanol can be substituted for the NuPAGE Sample Reducing Agent, with equivalent results. A final concentration of 2-5% beta-mercaptoethanol is usually sufficient to reduce the sample.

How much methanol do you recommend adding to the NuPAGE transfer buffer for transfer of NuPAGE Bis-Tris gels and NuPAGE Tris-Acetate gels?

We recommend adding 10% methanol to the NuPAGE transfer buffer for transfer of one gel and 20% methanol for the transfer of 2 gels.

Do you recommend adding the NuPAGE Antioxidant to the NuPAGE transfer buffer when I transfer proteins from NuPAGE Bis-Tris or NuPAGE Tris-Acetate gels?

Yes, we recommend adding the NuPAGE Antioxidant to the NuPAGE transfer buffer for enhanced blotting results with reduced proteins in order to maintain the reduced state of the proteins throughout the run.

What stains do you recommend for NuPAGE gels?

NuPAGE gels are compatible with any of the standard Coomassie staining procedures. The protocols that are accelerated by heat are preferable as the heat serves as a “fix” for proteins, especially smaller peptides. NuPAGE gels are also compatible with most silver staining protocols. They are also compatible with copper or zinc staining, and fluorescent stains.

Can I use NuPAGE Bis-Tris gels with NuPAGE MOPS or MES Running Buffer prepared without SDS for electrophoresis under native conditions?

We do not recommend using NuPAGE Bis-Tris gels with NuPAGE MOPS or MES Running Buffer prepared without SDS for electrophoresis under native conditions. This buffer system may generate excessive heat, resulting in poor band resolution. Further, the protein of interest may not migrate very well in a neutral pH environment if it is not charged.

How are Bolt Bis-Tris Plus gels different from NuPAGE Bis-Tris gels?

While they are both Bis-Tris based gels, the chemistries are very different as Bolt Bis-Tris Plus gels have been optimized for western blotting. Another key difference is the enabling wedge well design for Bolt Bis-Tris Plus gels, which allows larger sample volume load.

What are the main advantages of NuPAGE gels over Invitrogen Tris-Glycine gels?

NuPAGE gels have the following advantages over Tris-Glycine gels:

*Higher stability and longer shelf life: NuPAGE Bis-Tris gels and NuPAGE Tris-Acetate gels have a lower operating pH (pH 7 for NuPAGE Bis-Tris gels and pH 8.1 for NuPAGE Tris-Acetate gels) than Invitrogen Tris-Glycine gels (pH 9.5). At basic pH, polyacrylamide hydrolyzes to polyacrylic acid and ammonia whereas at neutral pH, this hydrolysis is slower. Hence, NuPAGE gels have higher stability and longer shelf life than Invitrogen Tris-Glycine gels (12 months at 4-25 degrees C for NuPAGE Bis-Tris gels and 8 months at 4 degrees C for NuPAGE Tris-Acetate gels vs 4-8 weeks at 4 degrees C for Tris-Glycine gels).

*Better resolution of proteins due to:

- Reduced undesired chemical modifications: Free acrylamide alkylates proteins at basic pH (8.5 to 9.0). It targets sulfhydryl cysteines and amine groups at the N-terminus and on lysines. This modification does not happen at pH below 8. Hence, proteins run on NuPAGE gels undergo fewer of these undesired chemical modifications than those run on Tris-Glycine gels.

- Reduced hydrolysis of proteins: Heating of Tris-Glycine sample buffer (pH 6.8) results in a drop in pH, causing Asp-Pro cleavage of proteins. High temperature and longer duration of heating/boiling increase the rate of this cleavage resulting in multiple peptide bands of decreased intensity. At 100 degrees C, the pH drops as low as pH 4.3. On the other hand, NuPAGE LDS sample buffer (pH 8.5) drops to pH 8.1 when heated to 70 degrees C, avoiding this cleavage.

*Faster run times: 35-50 min for NuPAGE Bis-Tris gels and 1 hour for NuPAGE Tris-Acetate gels vs 90 min for Tris-Glycine gels

How does the operating pH for Tris-Glycine gels differ from that for NuPAGE Bis Tris and NuPAGE Tris-Acetate gels?

The operating pH for Tris-Glycine gels is 9.5; the operating pH for NuPAGE Bis-Tris gels is 7 and for NuPAGE Tris-Acetate gels is 8.1.

How do you recommend transferring Midi gels?

Midi gels can be transferred using:

*iBlot Dry Blotting System in conjunction with Transfer Stacks
*Invitrogen Semi-Dry Blotter for simultaneous transfer of up to 2 Midi-gels
*Thermo Scientific Power Blotter for simultaneous transfer of up to 2 Midi gels
*Thermo Scientific G2 Fast Blotter (will be discontinued as soon as we exhaust current inventory).

Will NP-40 affect the migration of my protein samples?

All detergents, or even phospholipids in cell extracts, will form mixed micelles with SDS and migrate down into the gel. They can also interfere with the SDS:protein binding equilibrium. Most of the non-ionic detergents, including NP-40, are the worst at interfering with SDS-PAGE. The rule of thumb is to keep the ratio of SDS to lipid or other detergent at 10:1 or greater to minimize these effects.

Do your Invitrogen protein gels contain any carbohydrates and are they suitable for carbohydrate analysis?

All Invitrogen protein gels contain sucrose as a density-adjusting agent to facilitate pouring of the gel. Protein samples run on Invitrogen gels would be contaminated with large amounts of sucrose. Thus, Invitrogen gels are not recommended for this application.

What is the material used for making your Invitrogen precast gel plastic cassettes?

The cassettes are made of a styrene copolymer.

Can I recycle your Invitrogen precast gel plastic cassettes?

We do not recommend recycling our plastic cassettes because they have a chemical coating on them that may produce toxic fumes when melted and potentially cause contamination.

What is the difference between Invitrogen Mini and Midi gel formats?

Midi gels are wider than Mini gels and hence have a larger number of wells to accommodate additional samples in one gel. An experiment from a Mini gel can be easily scaled-up to a Midi gel of the same gel chemistry.

Midi gels:
*NuPAGE Bis-Tris, NuPAGE Tris-Acetate, & Invitrogen Tris-Glycine: Gel dimensions are 13cm x 8.3cm and Cassette dimensions are 15cm x 10.3cm.

Mini gels:
*NuPAGE Bis-Tris, NuPAGE Tris-Acetate, & Invitrogen Tris-Glycine: Gel dimensions are 8cm x 8cm and Cassette dimensions are 10cm x 10cm.
*New Bolt Bis-Tris Plus (Cat. No. NWxxxxxBOX): Gel dimensions are 8cm x 8.3cm and Cassette Dimensions are 10cm x10cm.
*Original Bolt Bis-Tris Plus (Cat. No. BGxxxxxBOX): Gel dimensions are 8cm x 8.3cm and Cassette Dimensions are 10cm x 10.5cm.

What are the dimensions of your precast protein gels?

All of our Invitrogen precast protein gels (NuPAGE gels, Bolt Bis-Tris Plus gels, and Novex gels) are available in Mini format. Our Mini gel dimensions are 8 cm x 8 cm and the cassette dimensions are 10 cm x 10 cm.

Our NuPAGE Bis-Tris, NuPAGE Tris-Acetate, and Novex Tris-Glycine Plus gels are also available in the wider Midi format. Our Midi gel dimensions are 8 cm x 13 cm and the cassette dimensions are 10 cm x 15 cm.

Are your precast protein gels available in Mini and Midi formats?

All our Invitrogen protein gels are available in Mini format. Certain gel chemistries (NuPAGE Bis-Tris, NuPAGE Tris-Acetate, and Invitrogen Tris-Glycine gels) are also available in the wide Midi format.

Note that Bolt Bis-Tris gels are not available in the Midi format and our Thermo Scientific Precise precast gels are only available in Mini format.

When running two protein gels, do I need to double the voltage?

If you are running the gels at constant voltage, you do not need to increase the voltage regardless of the number of gels. However, the resulting current and wattage observed will multiply linearly with the number of gels. Keep in mind that the expected total current for your gels should not exceed the current limit of the power supply, or else the current will plateau and the run will slow down. (For example: Recommended constant voltage for running a NuPAGE Bis-Tris gel with MES Buffer is 200 V, with a starting current of 110-125 mA/gel and end current of 70-80 mA/gel. If the power supply has a current limit of 500 mA, the maximum number of NuPAGE Bis-Tris gels that can be run at one time with full power is 500 mA/125 mA = 4 gels. Any additional gels will decrease the current per gel and increase the run time.

Can I run reduced and non-reduced protein samples on the same gel?

We do not recommend running reduced and non-reduced protein samples on the same gel, especially in adjacent lanes, since the reducing agent may have a carry-over effect on the non-reduced samples if they are in close proximity.

Can I store my reduced protein samples for later use?

We do not recommend storing reduced protein samples for long periods of time even if they are frozen because reoxidation of the sample may happen during storage, causing inconsistent results.

What is the ratio of acrylamide:bisacrylamide and percentage of cross-linker in your Invitrogen precast gels?

*Tris-Glycine gels (except 4% Tris-Glycine gels) have a 34.5:1 Acrylamide:bisacrylamide and 2.6% Crosslinker.

*4% Tris-Glycine gels have a 76:1 ratio Acrylamide:bisacrylamide and 1.3% Crosslinker.

What is the percentage of the stacking gel in your Invitrogen precast protein gels?

The percentage of the stacking gel is 4% in most of our gels including the Bolt Bis-Tris Plus gels. The NuPAGE Tris-Acetate gels contain a 3.2% stacking gel.

Do your Invitrogen precast protein gels contain a stacking gel?

Our Invitrogen precast protein gels contain a stacking gel that is ~8 to 9 mm long (it ends right above the first ridge on the cassette). The manufacturing method used results in an interface between the stacking and resolving gels that is not visually detectable.

What are the recommended sample loading volumes and protein loading amounts for your precast protein gels?

*Tris-Glycine and Invitrogen Tricine Mini gels: see here (http://tools.thermofisher.com/content/sfs/manuals/electrophoresisguide_man.pdf), Page 8

*NuPAGE Tris-Acetate and NuPAGE Bis-Tris Mini gels: see here (http://tools.thermofisher.com/content/sfs/manuals/nupage_tech_man.pdf), Page 10

*Bolt Bis-Tris Plus Mini gels: see here (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/bolt-bis-tris-gels.html)

*Thermo Scientific Precise Tris-HEPES gels: see here (https://tools.thermofisher.com/content/sfs/manuals/MAN0011499_Precise_Protein_Gels_UG.pdf), Page 1

*Midi gels (Invitrogen Tris-Glycine, NuPAGE Bis-Tris and NuPAGE Tris-Acetate): see here (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/novex_midigel_man.pdf), Page 4

*Thermo Scientific Precise Tris-Glycine gels: see here (https://tools.thermofisher.com/content/sfs/manuals/D25MAN0011814_Precise_TrisGlycine_Gels_UG.pdf), Page 1

Do your precast protein gels contain SDS?

Our precast protein gels do not contain SDS but they can be run under denaturing conditions when used with the appropriate denaturing running buffer.
Note: NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels, and Thermo Scientific Precise Tris-HEPES gels cannot be run under native conditions; they can only be run under denaturing conditions.

*Invitrogen Tris-Glycine gels: For Native electrophoresis, use Invitrogen Tris-Glycine Native Running Buffer. For Denaturing electrophoresis, use Invitrogen Tris-Glycine SDS Running Buffer

*NuPAGE Tris-Acetate gels: For Native electrophoresis, use Invitrogen Tris-Glycine Native Running Buffer. For Denaturing electrophoresis, use NuPAGE Tris-Acetate SDS Running Buffer

*NuPAGE Bis-Tris gels: For Denaturing electrophoresis, use NuPAGE MOPS-SDS Running Buffer or NuPAGE MES-SDS Running Buffer

*Bolt Bis-Tris Plus gels: For Denaturing electrophoresis, use Bolt MOPS SDS Running Buffer or Bolt MES SDS Running Buffer

*Thermo Scientific Precise Tris-Glycine gels: For Native electrophoresis, use Tris-Glycine SDS Running Buffer without SDS added. For Denaturing electrophoresis, use Tris-Glycine SDS Running Buffer.

*Thermo Scientific Precise Tris-HEPES gels: For Denaturing electrophoresis, use Tris-HEPES SDS Running Buffer.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.