missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Mini Protein Gels, 10%, 1.0–1.5 mm

Product Code. 10063412
Change view
Click to view available options
Wells:
1-well
10-well
12-well
15-well
2D-well
9-well
Gel Thickness:
1.0 mm
1.5 mm
Quantity:
10 gels (1 box)
2 gels (1 box)
Unit Size:
Each
Pack of 10
Pack of 2
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Gel Thickness Quantity unitSize
10063412 12-well 1.0 mm 10 gels (1 box) Each
10184862 10-well 1.0 mm 10 gels (1 box) Each
12807582 10-well 1.0 mm 2 gels (1 box) Each
12580137 12-well 1.0 mm 2 gels (1 box) Pack of 2
10398622 15-well 1.0 mm 10 gels (1 box) Each
12020156 1-well 1.0 mm 10 gels (1 box) Pack of 10
12030156 2D-well 1.0 mm 10 gels (1 box) Pack of 10
12040156 9-well 1.0 mm 10 gels (1 box) Pack of 10
12050156 10-well 1.5 mm 10 gels (1 box) Pack of 10
12060156 15-well 1.5 mm 10 gels (1 box) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10063412 Supplier Invitrogen™ Supplier No. NP0302BOX

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

One box of 10 gels

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Features of NuPAGE Bis-Tris gels:
Better protein integrity—optimized sample preparation process preserves your proteins
Wide ranges of molecular weight separation—select the right gel and running buffer to get the optimal separation of your proteins
Faster run times—get separation of your proteins in as little as 35 minutes
Longer shelf life—NuPAGE Bis-Tris gels can be stored for at least 12 months at room temperature

Choose the right NuPAGE Bis-Tris gel for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the Mini Blot Module (B1000) or XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB210010).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 10%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Recommended Applications Denaturing
Sample Loading Volume up to 20 μL
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 12-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Room Temperature
Storage Requirements Store at 4–25°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line NuPAGE
For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.