missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm

Product Code. 10277252
Change view
Click to view available options
Wells:
12 + 2-well
12 + 2-well, w/adapters
20-well
20-well, w/adapters
26-well
26-well, w/adapters
Unit Size:
1 set
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
10277252 26-well 1 set
10595013 12 + 2-well Each
10154702 12 + 2-well, w/adapters 1 set
10184482 20-well Each
10154912 20-well, w/adapters 1 set
10729243 26-well, w/adapters 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10277252 Supplier Invitrogen™ Supplier No. WG1603BOX

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

One box of 10 gels

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system.

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Features of NuPAGE Tris-Acetate gels:
High resolution—gels offer optimal separation of high molecular weight proteins
Better protein integrity—sample preparation process has been optimized to help preserve proteins
Longer self life—gels can be stored for at least eight months

Choose the right NuPAGE Tris-Acetate gel for your protein separation
Obtain optimal separation of your high molecular weight proteins by choosing the right combination of gel and running buffer. NuPAGE Tris-Acetate protein gels come in a polyacrylamide concentration of 7% and a 3–8% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Tris-Acetate gels also come in multiple well formats. Mini gels can be run using our XCell SureLock Mini-Cell (EI0001) or Mini Gel Tank (A25977). Midi gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or conveniently with the Bio-Rad Criterion™ Cell using our adapters (WA0999).

Run your proteins in native or denatured form
NuPAGE Tris-Acetate protein gels do not contain SDS and can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Tris-Acetate SDS Running Buffer (LA0041). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC26720.

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB210010).

TRUSTED_SUSTAINABILITY

Specifications

Adapters No Adapters
Gel Percentage 3 to 8%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Tris-Acetate
Recommended Applications Denaturing, Native
Sample Loading Volume Up to 15 μL
Separation Range 36 to 500 kDa
Separation Type Denaturing, Native
Wells 26-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Storage Requirements Store at 2°C to 8°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 13 cm
Product Line NuPAGE
For Use With (Equipment) XCell4 SureLock Midi-Cell
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.