missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Mini Protein Gels, 4–12%, 1.0–1.5 mm

Product Code. 12090156
Change view
Click to view available options
Wells:
1-well
10-well
12-well
15-well
17-well
2D-well
9-well
IPG-well
Gel Thickness:
1.0 mm
1.5 mm
Quantity:
10 gels (1 box)
2 gels (1 box)
Unit Size:
Each
Pack of 10
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Gel Thickness Quantity Unit Size
12090156 2D-well 1.0 mm 10 gels (1 box) Pack of 10
10472322 10-well 1.0 mm 10 gels (1 box) Each
12313623 10-well 1.0 mm 2 gels (1 box) Each
10247002 12-well 1.0 mm 10 gels (1 box) Each
12323623 12-well 1.0 mm 2 gels (1 box) Each
10338442 15-well 1.0 mm 10 gels (1 box) Each
12333623 15-well 1.0 mm 2 gels (1 box) Each
12080156 1-well 1.0 mm 10 gels (1 box) Pack of 10
12000166 9-well 1.0 mm 10 gels (1 box) Pack of 10
10780345 17-well 1.0 mm 10 gels (1 box) Each
12827582 17-well 1.0 mm 2 gels (1 box) Each
12010166 IPG-well 1.0 mm 10 gels (1 box) Pack of 10
12020166 10-well 1.5 mm 10 gels (1 box) Pack of 10
12343623 10-well 1.5 mm 2 gels (1 box) Each
12030166 15-well 1.5 mm 10 gels (1 box) Pack of 10
12353623 15-well 1.5 mm 2 gels (1 box) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 12090156 Supplier Invitrogen™ Supplier No. NP0326BOX
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Features of NuPAGE Bis-Tris gels:
Better protein integrity—optimized sample preparation process preserves your proteins
Wide ranges of molecular weight separation—select the right gel and running buffer to get the optimal separation of your proteins
Faster run times—get separation of your proteins in as little as 35 minutes
Longer shelf life—NuPAGE Bis-Tris gels can be stored for at least 12 months at room temperature

Choose the right NuPAGE Bis-Tris gel for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the Mini Blot Module (B1000) or XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 2D-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Room Temperature
Storage Requirements Store at 4–25°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line NuPAGE
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less
Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

What is the advantage of NuPAGE Gels over regular Tris-Glycine gels?

The neutral operating pH of the NuPAGE Gels and buffers provides following advantages over the Laemmli system:
-Longer shelf life of 8-12 months due to improved gel stability
-Improved protein stability during electrophoresis at neutral pH resulting in sharper band resolution and accurate results (Moos et al, 1998)
-Complete reduction of disulfides under mild heating conditions (70 degrees C for 10 min) and absence of cleavage of asp-pro bonds using the NuPAGE LDS Sample buffer (pH > 7.0 at 70 degrees C)
-Reduced state of the proteins maintained during electrophoresis and blotting of the proteins by the NuPAGE Antioxidant
Please refer to the following paper: Moos M Jr, Nguyen NY, Liu TY (1988) Reproducible High Yield Sequencing of Proteins Electrophoretically Separated and Transferred to an Inert Support. J Biol Chem 263:6005-6008.

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

Is it possible to run reduced and non-reduced samples on the same NuPAGE gel? Should the Antioxidant be used?

If the Antioxidant is omitted from the running buffer, it is possible to resolve reduced and non-reduced samples on the same gel, although the resolution may be lower. Furthermore, it is not recommended that the reduced and non-reduced samples be run side-by-side in adjacent lanes.
However, because of the neutral pH of the NuPAGE gels, the reducing agent (beta-mercaptoethanol or DTT) will not migrate through the gel with the protein the way it does in the basic environment of the Tris-Glycine gels. Instead, the reducing agent tends to remain at the top of the gel. For this reason, the NuPAGE Antioxidant is incorporated into the buffer in the upper buffer chamber. The antioxidant is able to migrate fully with the proteins and keep them reduced. As a result, it is possible that proteins prepared as non-reduced samples could become somewhat reduced during the electrophoresis run. This would result in smearing of the samples.

Can I use CTAB rather than SDS in my sample buffer?

No, CTAB will not work with any of our gels except for the NuPAGE Tris-Acetate gels. To use CTAB, you would need to use a running buffer of 50 mM acetic acid and 50 mM beta-alanine in equal concentrations. You would also need to switch the electrodes. Since CTAB is a cationic detergent, this would establish conditions for running a basic protein towards the anode (into the gel).

What is the length of the 2D well used with the precast gels?

The length of the 2D well is 6.5 cm. The IEF gel strip must be trimmed down to 5.8-5.9 cm before positioning it into the 2D well, even after it has been soaked in methanol to shrink the gel. For the 7 cm IPG strips, significant trimming would be necessary if the gels with the 2D wells are to be used. For IPG strips, the longer IPG well (found in the ZOOM gels) is recommended.

Does the NuPAGE 4 to 12%, Bis-Tris, 1.0–1.5 mm, Mini Protein Gels (Cat. No. NP0324BOX) have an extra well to load DNA ladder?

No. The 1-well NuPAGE 4 to 12%, Bis-Tris, 1.0–1.5 mm, Mini Protein Gels (Cat. No. NP0324BOX) only has one well. However, we do offer a 2D-well format of the (Cat. No. NP0326BOX) that has an additional well.

Do you have a protocol for extracting proteins from polyacrylamide gels?

We have a protocol for extracting proteins from polyacrylamide gels that provides a couple of options for identifying and excising the band of interest and eluting protein from gel matrix. Additionally, the article Extraction of Proteins from gels-a brief review by Kurien and Scofield (2020) provides an overview of elution procedures.

I ran my protein under native conditions on a Tris-Glycine gel. It has a pI that is higher than the pH of the Tris-Glycine transfer buffer. Can you offer some tips for transferring it?

- Increase the pH of Tris-Glycine transfer buffer to 9.2, allowing all the proteins below pI 9.2 to transfer towards the anode electrode.
- Use the Tris-Glycine transfer buffer and place a membrane on both sides of the gel. If there are any proteins that are more basic than the pH of the transfer buffer, they will be captured on the extra membrane placed on the cathode side of the gel. Both membranes can then be developed in the same manner.
- Prior to blotting, incubate the gel for 15 minutes in Tris-Glycine transfer buffer containing 0.1% SDS. The small amount of SDS will give the proteins enough charge to move unidirectionally towards the anode and in most cases, should not denature the protein. Proceed with the transfer using regular Tris-Glycine transfer buffer.

Do I need to add the chlorobutanol when making the 20X NuPAGE Transfer Buffer?

Chlorobutanol is used as a preservative in the NuPAGE transfer buffer and is not necessary for efficient transfer of proteins. You may prepare the buffer without chlorobutanol but keep in mind that the buffer will not be stable for long periods. We recommend using it within 2 weeks.

Can I transfer NuPAGE gels using Carbonate or CAPS transfer buffers?

We do not recommend using Carbonate or CAPS transfer buffers to transfer NuPAGE gels as the transfer efficiency will be badly compromised. Further, the high pH environment (>pH 9) of these buffers will make the NuPAGE Antioxidant non-functional.

Do you have any tips to improve transfer of high molecular weight proteins from NuPAGE gels?

To increase efficiency of transfer of high molecular weight proteins from NuPAGE gels, we recommend pre-equilibrating the gel in 2x NuPAGE Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1x NuPAGE transfer buffer containing methanol and 0.01% SDS.

My gel run is faster than normal with poor resolution. What could be causing this problem?

Here are possible causes and solutions:

- Buffers are too concentrated or incorrect. Check buffer recipe; dilute or re-make if necessary.
- Voltage, current or wattage is set at a higher limit. Decrease power conditions to recommended running conditions.

My gel will not run. Can you please help me troubleshoot?

It is important to determine whether the problem is with the power supply, the apparatus or the gel. Often, it helps to switch out the power supply or the lid to see if there is a faulty contact. Also, check to see whether the tape from the bottom of the gel cassette has been removed and whether the buffer core is damaged. Additionally, make sure there is sufficient buffer in the electrophoresis tank to cover the wells of the gel.

Which of your protein gels can I run using the Mini Gel Tank?

Our New Bolt Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Invitrogen Mini gels and NuPAGE Mini gels can be run using the Mini Gel Tank. Please note that our original Bolt Bis-Tris Plus Mini gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) can only be run in the Bolt Mini Gel Tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).

Which of your protein gels can I run using the XCell SureLock Mini-Cell?

Our New Bolt Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Invitrogen Mini gels and NuPAGE Mini gels can be run using the XCell SureLock Mini-Cell.

After western detection, my membrane has a lot of spots. What could have gone wrong?

Here are possible causes and solutions:

- Membrane blotting pads are dirty or contaminated. Soak pads with detergent and rinse thoroughly with purified water before use. Replace pads when they become worn or discolored.
- Blocking was uneven. The incubation dish must be sufficiently big to allow thorough coverage of membrane. Shake or agitate during each step.

I am getting a lot of non-specific binding after western detection. Can you offer some tips?

Here are possible causes and solutions:

- Membrane contaminated by fingerprints or keratin proteins: Wear clean gloves at all times and use forceps when handling membranes. Always handle membranes around the edges.
- Concentrated secondary antibody used: Make sure the secondary antibody is diluted as recommended. If the background remains high, but with strong band intensity, decrease the concentration of the secondary antibody.
- Concentrated Primary antibody used: Decrease the concentration of the primary antibody.
- Affinity of the primary antibody for the protein standards: Check with the protein standard manufacturer for homologies with primary antibody.
- Insufficient removal of SDS or weakly bound proteins from membrane after blotting: Follow instructions for membrane preparation before immunodetection.
- Short blocking time or long washing time: Make sure that each step is performed for the specified amount of time.

I am getting very high background after western detection. Can you please offer some tips?

Here are possible causes and solutions:

- Insufficient blocking or non-specific binding: We suggest trying our WesternBreeze Blocker/Diluent (Cat. No. WB7050).
- Membrane is contaminated: Use only clean, new membranes. Wear clean gloves at all times and use forceps when handling membranes.
- Higher intrinsic background with PVDF membranes: Switch to nitrocellulose membranes.
- Nitrocellulose membrane not completely wetted: Follow instructions for pre-wetting the membrane.
- Blot is overdeveloped: Follow recommended developing time and remove blot from substrate when signal - to -noise ratio is acceptable.
- Insufficient washing ; Follow recommended number of washes. In some cases, it may be necessary to increase the number or duration of washes.
- Concentrated secondary antibody used: Determine optimal antibody concentration by performing a dot blot and dilute antibody as necessary.
- Concentrated primary antibody used: Determine optimal antibody concentration by performing a dot blot and dilute antibody as necessary.

I am unable to visualize my protein bands after western detection. What is the problem?

Here are possible causes and solutions:

- The primary antibody and secondary antibody are not compatible: Use a secondary antibody that was raised against the species in which the primary antibody was raised.
- The primary antibody is too dilute: 1) Use a more concentrated antibody solution. 2) Incubate longer (e.g., overnight) at 4 degrees C. 3) Use fresh antibody and keep in mind that each time an antibody solution is used, its effective antibody concentration decreases.
- Something in your blocking buffer interferes with binding of the primary and/or secondary antibody: Try an alternate blocking buffer ± a mild surfactant like Tween-20 (0.01-0.05% v/v). There are many blocking buffer recipes available, based on non-fat dry milk, BSA, normal serum, gelatin and mixtures of these and other materials. Note that BSA (1-5%) is considered the best blocker for nitrocellulose membranes. It is easy to check the efficacy of different blocking buffers by performing dot-blots.
- The primary antibody does not recognize the protein in the species being tested: 1) Evaluate primary antibodies by dot-blotting first to how well they react with your protein. 2) Check the immunogen sequence, if provided, and determine if it is found in your protein. 3) If no immunogen sequence is available, perform a PubMed/BLAST alignment to assess the degree of homology between your target protein and the protein against which the antibody was generated. Note that many antibodies against human proteins will also recognize the non-human primate version because there is usually a high degree of amino acid identity. In contrast, many antibodies against human proteins will not recognize the corresponding proteins from rodents (and vice versa). Remember that significant homology between sequences does not guarantee that the antibody will recognize your protein. 4) Always run the recommended positive control, if available.
- Insufficient protein is bound to the membrane or the protein of interest is not abundant enough in the sample: 1) Load at least 20-30 ?g protein per lane on your gels (as a starting point), since proteins representing less than ~0.2% of the total protein are difficult to detect on western blots. 2) Use an enrichment step to increase the concentration of the target protein. For example, prepare two nuclear lysates prior to blotting nuclear proteins or perform an immunoprecipitation (IP) prior to SDS-PAGE. 3) Reduce the volume of cell extraction buffer used to lyse your cells or tissue. 4) Be sure to use freshly prepared protease inhibitors and phosphatase inhibitors, if needed, in your protein extraction buffer. 5) Run the recommended positive control, if available.
- Poor or no transfer of the proteins to the membrane 1) Check the protein transfer efficiency with a reversible protein stain like Invitrogen Reversible Membrane Protein Stain, ponceau S, amido black or use pre-stained molecular weight standards. 2) Verify that the transfer was performed with the correct electrical polarity. 3) Remember that proteins with basic pI values (e.g., histones) and high MW may not transfer well. 4) Remember that if your target protein has a low MW (≤10 kDa), it may transfer more quickly than expected. 5) If you are using PVDF membranes, make sure to pre-soak the membrane in methanol first before soaking it in transfer buffer. Note that methanol in transfer buffer increases protein binding to nitrocellulose, but omitting methanol can increase transfer efficiency of high MW proteins. 6) Low MW proteins may pass through the 0.45 µm pores in nitrocellulose membranes, so switch to NC with 0.2 or 0.1 µm pores instead.
- Excessive washing or blocking of the membrane:- 1) Avoid over-washing the membrane. Extra washing will not allow you to visualize your protein of interest if there are other problems with your blot. 2) Avoid over-blocking by using high concentrations of the blocking buffer components or long incubation times. Too much blocking can prevent your antibodies from binding to your protein. Gelatin, in particular, can mask proteins on the blot, so avoid it, if possible. Milk can also mask proteins, so instead of using 5% milk in your blocking buffer, try using it at 0.5% instead, or remove it altogether. 3) Switch to a different blocking reagent and/or block the blot for less time.
- Using the same solution of diluted primary antibody repeatedly: Use freshly-diluted antibody for each western blot because the effective concentration of a diluted antibody decreases each time it is re-used. Also, remember that dilute solutions of antibodies are less stable and may lose their activity rapidly.
- The enzyme conjugated to your secondary antibody is not working: 1) Make a fresh dilution of your secondary antibody conjugate each time you need it. Enzymes (and antibodies) may lose activity quickly in dilute solutions. 2) Omit sodium azide in buffers if you are using HRP-conjugated antibodies. 3) Avoid high heme concentrations (from blood contamination), which can interfere with HRP-based detection. 4) Avoid using phosphate in buffers with alkaline phosphatase-antibody conjugates because phosphate inhibits enzyme activity.
- Your colorimetric or other detection reagent is old and inactive: 1) Use fresh enzyme substrate for each experiment. 2) Don't use ready-to-use substrate reagents if they have changed color on their own or if they have passed their expiration date. 3) Do not dilute substrate solutions unless instructed to do so in the product manual.

I ran my protein sample on one of your gels and the bands look non-distinct and smeary after western detection. What should I do?

Here are some suggestions:

- Make sure that the correct amount of protein is loaded per lane; loading too much protein can cause smearing.
- Bands will not be as well resolved in low percentage gels; try using a higher percentage gel.
- This may be due to the antibody being too concentrated. We recommend following the manufacturer's recommended dilution or determining the optimal antibody concentration

I stained my 2D gels and got uneven background staining. Can you please offer some tips?

Here are the possible causes and solutions:
*Staining protocol modified; Refer to the manufacturer's recommendations for correct staining protocols. To achieve best results, be sure to follow all steps exactly as given in the protocol, especially for silver staining. Changes in the protocol can result in high background.
*Background staining due to ampholytes: Prior to staining the 2D gel, thoroughly wash the gel to remove ampholytes. Use ZOOM Carrier Ampholytes as they provide a clear background due to very low non-specific binding of dyes and stains.

Can I heat my protein sample prepared using ZOOM 2D Protein Solubilizer?

We do not recommend heating protein samples containing urea over 37 degrees C as elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.

How do you recommend storing protein samples prepared in the Sample Rehydration buffer?

We recommend storing them at -80 degrees C. We do not recommend storing them at -20 degrees C

How much ampholyte do you recommend adding to the sample rehydration buffer?

The recommended ampholyte concentration in the sample rehydration buffer is 0.5%.

*If you are loading 5-50 µg of protein (pure protein or crude lysate) per ZOOM strip, use 0.5% ampholytes in the sample rehydration buffer.
*If you are loading >50 µg of protein (crude lysate or fractionated sample) per ZOOM Strip, use 0.5-2% ampholytes in the sample rehydration buffer.

Note: Higher ampholyte concentration requires longer focusing times.

What is the maximum recommended volume of protein sample to be added to the Sample Rehydration buffer?

The maximum volume of the protein sample should at most be 1/6 of the final sample volume that will be added to the strip. A good general volume would be 5-10µL. 140 µL of sample diluted in Sample Rehydration buffer is used to rehydrate each ZOOM Strip for the standard rehydration time of one hour. For overnight rehydration, we recommend using 155 µL of diluted sample.

What is the purpose of alkylation prior to IEF?

Alkylation prevents unwanted protein modifications by alkylating cysteines to avoid mixed disulfide formation and reoxidation and this allows for crisper focusing.

Do you still offer ZOOM 2D Protein Solubilizers?

Sorry we have discontinued selling the ZOOM 2D Protein Solubilizers.

What are the main reasons for performing 2D gel electrophoresis of proteins?

The main advantages of performing 2D gel electrophoresis of proteins and applications used for are listed below:

Advantages:

*Simultaneous separation of hundreds to thousands of proteins
*High capacity with superior resolution
*Compatible with further analysis by MS for protein identification and sequencing
Ability to separate and analyze low-abundance proteins

Applications:

*Comparative proteomics: identifying and analyzing differences between complex mixtures of proteins
*Protein profiling, biomarker discovery
*Separation and analysis of protein variants and isoforms

What products do you offer for 2D gel electrophoresis of proteins?

We offer the following products for the first- and second-dimension separation of proteins:

First-dimension separation:

*Vertical gels for separation of proteins based on their isoelectric point (pI) (https://www.thermofisher.com/us/en/home/life-science/protein-expression-and-analysis/protein-gel-electrophoresis/protein-gels/specialized-protein-separation/isoelectric-focusing.html)

*Solution-phase isoelectric focusing of proteins using ZOOM Disks (immobilized buffer disks of specific pH) to reduce sample complexity, enrich low-abundance proteins, and increase the dynamic range of detection (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/isoelectric-focusing/zoom-ief-fractionator.html)

*Mini gel system for high-throughput isoelectric focusing of proteins using ZOOM IPG (Immobilized pH Gradient) Strips (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/2d-gel-electrophoresis/zoom-ipgrunner-system.html)

Second-dimension separation:

*ZOOM gels for 2D electrophoresis: NuPAGE Bis-Tris (Cat. No. NP0330BOX) and Tris-Glycine (Cat. No. EC60261BOX) mini gels with IPG wells ( to accommodate 7 cm ZOOM strips) for separation of proteins based on their molecular weight

What is 2D protein gel electrophoresis?

2D protein gel electrophoresis is the separation of proteins in two dimensions. In the first dimension, proteins are separated by their isoelectric point (pI) using isoelectric focusing, and in the second dimension, they are separated by their mass using SDS-PAGE.

The protein bands in some of my gel lanes are irregular or wavy? What would have caused this problem?

This could be due to:

*Debris in the well
*High salt in the sample (make sure that the salt concentration does not exceed 50-100 mM)
*Running buffer issue
*Gel casting error

I am seeing a very wavy and uneven dye front with my samples. Can you please help me troubleshoot?

This could be due to a gel polymerization issue combined with incorrect sample preparation (final sample dilution less than 1X). Please try a different lot of the same gel and make sure that the sample is correctly prepared.

I am seeing a faint, artifact doublet band at ~60 kDa in all my lanes. This band seems to be getting darker the longer I stain the gel. What could be causing this?

Possible cause:

*Excess reducing agent (beta-mercaptoethanol)
*Skin protein contaminants (keratin)

Remedy:

*The addition of iodoacetamide to the equilibration buffer just before applying the sample to the gel has been shown to eliminate these artifact bands.
*Use new electrophoretic solutions and wear gloves when handling and loading the gel. This issue is more common when highly sensitive stains are used.

I loaded different protein samples in each well but I see the same protein band in several neighboring lanes. What could have happened?

Possible cause:

*Carry-over contamination of sample from one well into neighboring wells due to loading error
*Contaminated running buffer
*Gel casting error: malformed wells

Remedy:

*Use a gel loading tip to load wells
*Reduce the sample volume
*Do not delay while loading wells
*Do not delay after the run, as proteins can diffuse horizontally; a full well left next to an empty well would eventually contaminate the empty well over time.

My protein bands appear to be skewed or distorted. What is the problem?

Possible cause:

*Poor polymerization around sample wells
*High salt concentration in sample
*Uneven gel interface
*Excessive pressure applied to the gel plates when the gel is placed into the clamp assembly
*Uneven heating of the gel
*Insoluble material in the gel or inconsistent pore size throughout the gel
*Air bubble during the run

Remedy:

*Remove excess salt/other material by dialysis, Sephadex G-25 or any other desalting column or using an Amicon concentrator.
*Either use a cooled apparatus or reduce the current at which electrophoresis is performed.

I ran my reduced protein samples under denaturing conditions and am seeing doublet protein bands when I expect to see single bands. Why is this happening?

A portion of the protein sample may have re-oxidized during the run, or may not have been fully reduced prior to the run. We recommend preparing fresh sample solution using fresh beta-mercaptoethanol or dithiothreitol (DTT). For NuPAGE gels, we recommend adding antioxidant to the running buffer.

My gel seems to be lifting off the cassette. What could be causing this?

Gel lifting off the cassette can be caused by:

*Expired gels that are degrading
*Improper storage of gels
*Too much heat accumulating during the electrophoresis run due to excessive current
*Insufficient polymerization of the polyacrylamide

I am seeing a faint shadow, or "ghost" band below a normal and expected protein band? What could be the potential issue?

Ghost bands are usually attributed to a slight lifting of the gel from the cassette, which results in the trickling down of some sample beyond its normal migration point. It then accumulates and appears as a faint second band.

My protein bands in the outer lanes of the gel show a "smiling" effect. Can you please help me troubleshoot?

"Smiling" bands may be the result of the acrylamide in the gel breaking down, leaving less of a matrix for the proteins to migrate. We recommend checking to ensure that the gels have not been used past their expiration date.

I see dumbbell or barbell shaped bands after protein electrophoresis. What could be causing this?

Barbell shaped bands are a result of loading too large of a sample volume. When a large sample volume is loaded, part of the sample tends to diffuse to the sides of the wells. When the run begins and the sample moves through the stacking portion of the gel, the sample will incompletely stack causing a slight retardation of the portion of the sample that diffused to the sides of the wells. This effect may be intensified for larger proteins, whose migration is more impeded in the low concentration acrylamide of the stacking gel. To alleviate the problem, we recommend concentrating the protein and loading a smaller volume. This gives a "thinner" starting zone.

Why do I get streaking forward or "frowning" of one of my samples on my protein gel?

Here are possible causes and solutions:

1) Sample overload: Do not overload samples
2) Addition of reducing agent that is not fresh: Reduce samples right before loading and do not use samples that have been stored in reducing agent
3)Re-oxidation of the protein during the run: Add antioxidant to the running buffer if you are running NuPAGE gels
4) Presence of highly hydrophobic regions where the protein can exclude SDS: Load the sample with 2X sample buffer instead of 1X sample buffer
5) Excess salt in the sample: Precipitate and reconstitute in lower salt buffer
6) Not enough SDS in the sample: Add SDS to the upper buffer chamber (try 0.1%, 0.2%, 0.3% and 0.4% SDS)


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.