missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Welcome Pack, 4 to 12%

Product Code. p-7224320
Change view
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Unit Size
15771778 10-well 1 set
15853561 12-well 1 set
15824052 15-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15771778 Supplier Invitrogen™ Supplier No. NP032A
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the Mini Gel Tank.

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

The NuPAGE Bis-Tris Welcome Pack contains:

  • Mini Gel Tank (A25977)
  • NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
  • NuPAGE MES SDS Running Buffer, 20X (NP0002)
  • MES and MOPS Running Buffer, Powder (NP000105, NP000205)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the Mini Gel Tank

The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Bis-Tris gels

Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Warranty

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Separation Range 3.5 to 260 kDa
Wells 10-well
Quantity 1 Welcome Pk. kit
Shipping Condition Approved for shipment at room temperature and wet ice
Product Line NuPAGE
For Use With (Equipment) Mini Gel Tank
I used one of your protein standards for a western transfer and noticed that some of the lower-molecular weight protein bands passed through the membrane. How can I resolve this issue?

- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.

I used one of your protein standards for a western transfer and noticed that some of the higher-molecular weight bands transferred very poorly to the membrane. Can you offer some tips?

- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.

I used one of your pre-stained standards on a Tris-Glycine gel and noticed that the molecular weights of the proteins were different than on a NuPAGE Bis-Tris gel. What is the reason for this?

Pre-stained standards have a dye that is covalently bound to each protein that will result in the standard migrating differently in different buffer systems (i.e., different gels). As a result, using a pre-stained standard for molecular weight estimation will only give the apparent molecular weight of the protein. Pre-stained standards may be used for molecular weight approximation, confirming gel migration and estimating blotting efficiency but for accurate molecular weight estimation, an unstained standard should be used.

I used one of your protein standards and am seeing some extra bands in the lane. Can you offer some suggestions?

- While loading, take care to make sure that there is no cross-contamination from adjacent sample lanes.
- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can result in extra bands and this is a problem especially with silver-stained gels.
- Improper storage of the standard or repeated freeze/thawing can result in protein degradation.

I used one of your protein standards and the bands look non-distinct and smeary. What should I do?

Here are some suggestions:

- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can cause smearing and this is a problem especially with silver stained gels.
- Bands will not be as well resolved in low percentage gels. Try using a higher percentage gel.
- If the bands look smeary and non-distinct after a western transfer/detection, this may be due to the antibody being too concentrated. Follow the manufacturer's recommended dilution or determine the optimal antibody concentration by dot-blotting.

A couple of bands in my protein standard are missing on the gel. Can you help me troubleshoot?

Here are some suggestions:

- Check the gel type/percentage of the gel that was used. Depending on the gel type and/or percentage, all the bands may not be seen. For example, the smallest bands of the protein standard may not resolve on a very low percentage gel whereas the higher molecular weight bands may not resolve on a high percentage gel.
- Check the expiration date on the protein standard. Expired lots may result in faded or missing bands due to protein degradation.
- Check the storage conditions for the protein standard. Improper storage conditions will compromise the stability of the proteins in the standard.
- Make sure that the protein standard was not heated/boiled prior to loading on the gel. Our protein standards are ready to load and we do not recommend heating/boiling them as this may cause degradation of proteins in the standard.

What is the composition of the PageRuler Plus Prestained Protein Ladder?

The PageRuler Plus Prestained Protein Ladder is a mixture of nine (9) blue-, orange- and green-stained proteins (10 to 250 kDa) for use in protein electrophoresis (SDS-PAGE) and western blotting. Two orange reference bands at ~70 kDa and 25 kDa and one green reference band at 10 kDa highlight the blue-stained protein ladder.

What are the storage conditions and shelf life for the PageRuler Prestained Protein Ladder and PageRuler Plus Prestained Protein Ladder?

We recommend storing these ladders at -20 degrees C where they are stable for a year. They are stable for up to 3 months at 4 degrees C.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.