missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NuPAGE™ Bis-Tris XCell SureLock™ Mini-Cell Welcome Pack, 4 to 12%

Product Code. 17759893 Shop All Thermo Scientific Products
Change view
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Unit Size
17759893 15-well Each
17739893 10-well Each
17749893 12-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17759893 Supplier Thermo Scientific™ Supplier No. NP032F
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.

The NuPAGE Bis-Tris Welcome Pack contains:

  • XCell SureLock Mini-Cell (EI0001)
  • NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
  • NuPAGE MES SDS Running Buffer, 20X (NP0002)
  • MES and MOPS Running Buffer, Powder (NP000105, NP000205)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the XCell SureLock Mini-Cell

The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Bis-Tris gels

Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Separation Range 3.5 to 260 kDa
Wells 15-well
Quantity 1 Welcome Pk.
Shipping Condition Approved for shipment at room temperature and wet ice
Product Line NuPAGE
For Use With (Equipment) XCell SureLock Mini-Cell

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.