Learn More
Invitrogen™ NuPAGE™ MES SDS Running Buffer (Powder)
Description
Invitrogen NuPAGE MES SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to enable enfast, high-resolution SDS-PAGE protein separations. The MES buffer system supports rapid protein migration and is recommended for resolving small- to medium-sized proteins. Supplied as a dry powder, this running buffer enables flexible preparation, room temperature storage, a two-year shelf life, and is designed to be a convenient and reliable choice for routine electrophoresis workflows.
Features and benefits include:
- Fast separations for small- to medium-sized proteins—MES supports faster protein migration than MOPS-based buffers, helping reduce run times while maintaining resolution
- Optimized for Bis-Tris gel workflows—designed for use with Bis-Tris gels including both NuPAGE Bis-Tris and Bolt Bis-Tris Plus gels
- Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify storage
- Room temperature storage and extended shelf life—powder format supports convenient storage with a two-year shelf life for inventory efficiency
- Reliable performance for routine SDS-PAGE—supports consistent results across common protein electrophoresis workflows when used under recommended conditions
Specifikationer
Specifikationer
| Color | White |
| Format | Powder |
| Gel Type | Bis-Tris |
| Formulation | MES SDS Running Buffer |
| For Use With (Application) | SDS-PAGE |
| Quantity | 5 Packets |
| Storage Requirements | Room Temperature |
| Sufficient For | Preparing 5 L Running Buffer |
| Product Line | NuPAGE |
| Product Type | SDS Running Buffer |
| Visa mer |
For Research Use Only. Not for use in diagnostic procedures.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.