missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (Powder)

Product Code. 31904851
Change view
Click to view available options
Quantity:
5 Packets
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31904851 5 Packets Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31904851 Supplier Invitrogen™ Supplier No. NP000205
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen NuPAGE MOPS SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to resolve medium- to large-sized proteins by providing slower SDS-PAGE separations. Supplied as a powder for flexible preparation, room temperature storage, and a long 2-year shelf life.

Invitrogen NuPAGE MOPS SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to improve the resolution and band sharpness of medium- to large-sized proteins. Compared to MES-based buffers, the MOPS buffer system supports reduced protein migration rates for enhanced separation performance. Supplied as a dry powder, this running buffer enables flexible preparation, room temperature storage, a two-year shelf life, and is designed to be a convenient and reliable choice for routine electrophoresis workflows.

Features and benefits include:

  • Improved resolution of medium- to large-sized proteins—slower protein migration with MOPS buffer enhances separation and band sharpness compared to faster MES-based buffers
  • Optimized for Bis-Tris gel workflows—designed for use with Bis-Tris gels including both NuPAGE Bis-Tris and Bolt Bis-Tris Plus gels
  • Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify laboratory workflows
  • Room temperature storage and extended shelf life—powder format enables convenient storage with a two-year shelf life for inventory efficiency
  • Reliable performance for routine SDS-PAGE—supports consistent and reproducible results across common protein electrophoresis applications when used under recommended conditions
TRUSTED_SUSTAINABILITY

Specifications

Color White
Format Powder
Gel Type Bis-Tris
Formulation MOPS SDS Running Buffer
For Use With (Application) SDS-PAGE
Quantity 5 Packets
Storage Requirements Room Temperature
Sufficient For Preparing 5 L Running Buffer
Product Line NuPAGE
Product Type SDS Running Buffer
For Use With (Equipment) Electrophoresis Chambers
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.