missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm

Product Code. 11541216
Change view
Click to view available options
Wells:
20-well
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
11541216 20-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11541216 Supplier Invitrogen™ Supplier No. WG1602BX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system.

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Invitrogen NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. They are high performance polyacrylamide gels that simulate the denaturing or native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of high molecular weight proteins compared to traditional Tris-glycine SDS-PAGE gels.

Features of NuPAGE Tris-Acetate gels include:

  • High resolution—gels offer optimal separation of high molecular weight proteins
  • Better protein integrity—sample preparation process optimized to help preserve proteins
  • Longer self life—gels can be stored for at least eight months

Choose the right NuPAGE Tris-Acetate gel for your protein separation

Obtain optimal separation of your high molecular weight proteins by choosing the right combination of gel and running buffer. NuPAGE Tris-Acetate protein gels come in a polyacrylamide concentration of 7% and a 3–8% gradient. Gels come in two sizes: mini (8 x 8 cm) or midi (8.7 x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Tris-Acetate gels also come in multiple well formats. Mini gels can be run using our XCell SureLock Mini-Cell (Cat. No. EI0001) or Mini Gel Tank (A25977). Midi gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or with the Bio-Rad Criterion™ Cell using our adapters (WA0999).

Run your proteins in native or denatured form

NuPAGE Tris-Acetate protein gels do not contain SDS and can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Tris-Acetate SDS Running Buffer (LA0041). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC26720).

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP00061) for traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C. Do not freeze.
Gel Percentage 3 to 8%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Tris-Acetate
Sample Loading Volume Up to 25 μL
Separation Range 36 to 500 kDa
Separation Type Denaturing, Native
Wells 20-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Length (Metric) 8 cm
Width (Metric) 13 cm
Product Line NuPAGE
For Use With (Equipment) SureLock Tandem Midi Gel Tank
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.