missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NuPAGE™ Tris-Acetate XCell SureLock™ Mini-Cell Welcome Pack, 3-8%

Product Code. p-7224380 Shop All Thermo Scientific Products
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17787483

Brand: Thermo Scientific™ EA0375D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.

The NuPAGE Tris-Acetate Welcome Pack contains:
• XCell SureLock Mini-Cell (EI0001)
• NuPAGE Tris-Acetate mini gels (2 boxes, 20 gels)
• NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
• NuPAGE LDS Sample Buffer, 4X (NP0007)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• HiMark Pre-stained Protein Standard (LC5699)

About the XCell SureLock Mini-Cell 
The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Tris-Acetate gels
NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer buffer (NP0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) XCell SureLock Mini-Cell
Gel Percentage 3 to 8%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Acetate
Separation Range 40 to 500 kDa
Wells 10-well
Product Line NuPAGE
Quantity 1 Welcome Pk.
Shipping Condition Approved for shipment at room temperature and wet ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.