missing translation for 'onlineSavingsMsg'
Learn More

Nutriacholic Acid, TRC

Product Code. 30012837
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012837 10 g 10g
30009812 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012837 Supplier Toronto Research Chemicals Supplier No. TRCN92555010G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4651-67-6
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym 3alpha-Hydroxy-7-oxo-5beta-cholan-24-oic Acid, 3alpha-Hydroxy-7-oxo-5beta-cholanic Acid, 3alpha-Hydroxy-7-oxo-5beta-cholanoic Acid, 3alpha-Hydroxy-7-oxo-5beta-cholic Acid, 5beta-Cholanic Acid-3alpha-ol-7-one, 7-Ketochenodeoxycholic Acid, 7-Ketolithocholic Acid, 7-Oxo-3alpha-hydroxycholan-24-oic Acid, 7-Oxolithocholic Acid, 7-Keto-Lithocholic Acid, NSC 226118, 3alpha-Hydroxy-7-oxo-5beta-cholanic Acid, 3alpha-Hydroxy-7-oxo-5beta-cholan-24-oic Acid, (3alpha,5beta)-3-Hydroxy-7-oxo-cholan-24-oic Acid, Ursodeoxycholic Acid EP Impurity F;
IUPAC Name (4R)-4-[(3R,5S,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-7-oxo-1,2,3,4,5,6,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@H]3[C@H](CC[C@]12C)[C@@]4(C)CC[C@@H](O)C[C@H]4CC3=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C24H38O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-19,22,25H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,22+,23+,24-/m1/s1
Formula Weight 390.277
Chemical Name or Material Nutriacholic Acid (3alpha-Hydroxy-7-oxo-5Beta-cholan-24-oic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.