missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVS-STG2
Description
NVS-STG2 is a STING agonist. It induces oligomerization of human STING by binding to pockets between adjacent STING dimer transmembrane domains, acting as a molecular glue. NVS-STG2 elicits potent STING-mediated immune responses in cells and produces antitumor activities in hSTING knock-in mice.
Specifications
Specifications
| Chemical Name or Material | 4-((4-(2-(tert-Butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoic acid |
| CAS | 3030588-01-0 |
| Quantity | 50 mg |
| Target | STING Agonists |
| Molecular Formula | C25H33NO5 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?