Learn More
Masterflex™ Nylon Clamps for Sanitary Fittings
Product Code. 13188169
Change view
Click to view available options
Width (Metric):
12.7 to 19.05 mm
25.4 to 38.1 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
13188169 12.7 to 19.05 mm Each
13198169 25.4 to 38.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13188169 Supplier Masterflex™ Supplier No. 3050821
Only null left
Add to basket
Add to basket
Maintain sanitary conditions with these clamps. Cole-Parmer™ Nylon Clamp for Sanitary Fittings feature excellent chemical resistance and low leachable characteristics.
  • Low leachable characteristics
  • Sterilize with Ethylene Oxide (ETO)
  • Compatible with Tri-Clamp™ fittings
Specifications
Material Nylon
For Use With Tri-Clamp™ Fitting
Packaging Quantity 1 Ea.
Width (Metric) 12.7 to 19.05 mm
Product Type Tubing Clamp