Learn More
Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves
Product Code. p-8010803
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
1 pair
12 pairs
Each
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12961135
Brand: Ansell Edmont™ C357100
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Providing good resistance to abrasion and small blade cut hazards. Ansell Edmont™ Marigold™ Picolon Confort™ Polyamide Gloves are ideal for the handling and assembly of slightly sharp and slippery objects.
  • Dots on palm and fingers for better grip
  • Textile liner made of high elasticity polyamide thread
  • Fine, stretchable and comfortable stitches
  • Knitted with a seamless non-stop process
  • Lint-free
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material PVC
Color White with Blue Dots
Size X-Large
Glove Cuff Style Knit
Liner Material Polyamide
Resistant To Abrasion and small cuts
Length (Metric) 260 mm