missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nylon Membrane Filter

Product Code. 11735315 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Diameter (Metric):
25 mm
47 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) unitSize
11735315 0.22 μm 25 mm Pack of 100
11745315 0.22 μm 47 mm Pack of 100
11755315 0.45 μm 25 mm Pack of 100
11765315 0.45 μm 47 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11735315
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Add to basket

Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.

  • Naturally hydrophilic membrane
  • Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
  • Flexible, durable and tear resistant
  • Autoclaved at 135°C
  • Effective chemical compatibility with aqueous and alcoholic solutions and solvents

Specifications

Autoclavable Autoclavable
Bubble Point 3.4bar
Color White
Filter Type Microfilter
Material Nylon
Pore Size 0.22 μm
Sterility Non-sterile
Flow Rate 9.9mL/min./cm2
For Use With (Application) Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs)
Gas Flow Rate 1.7L/min./cm2
Hydrophobicity Hydrophilic
Quantity 100 Pack
Type Plain Disc
Wettability Yes
Diameter (Metric) 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.