missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Nylon Ties

Product Code. 11796885 Shop All Brady Products
Change view
Click to view available options
Color:
Blue
Orange
Purple
Red
White
Yellow
Unit Size:
1 pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11796885 Red 1 pack
11776885 White 1 pack
11706895 Orange 1 pack
11786885 Blue 1 pack
11716895 Yellow 1 pack
11736895 Purple 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11796885 Supplier Brady™ Supplier No. 229957

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Cross-laminated polyethylene
  • Matte finish

Specifications

Type Ties
Material Nylon
Color Red
Width (Metric) 3.6 mm
Quantity 100/Pk.
For Use With (Application) Ideal for color-coding hard to mark items
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.