missing translation for 'onlineSavingsMsg'
Learn More

Nystatin, >70% A3, TRC

Product Code. 30007060
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007060 10 g 10g
17977324 25 g 25g
17979834 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007060 Supplier Toronto Research Chemicals Supplier No. TRCN92600010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1400-61-9
Molecular Formula C47H75NO17
Molecular Weight (g/mol) 926.09
Synonym Nystatin, Biofanal, Candio-Hermal, Ceratocide, Diastatin, Flagystatin, Fungicidin, L-Nystatin, Moronal (antibiotic), Myconystatin, Mycostatin, Mycostatin Pastilles, Mykostatyna, Nistatin, Nyotran, Nysfungin, Nystacid, Nystan, Nystavescent, Nystex, Nystop, O-V Statin, Stamycin
IUPAC Name Nystatin
SMILES O=C1O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)/C=C/C=C/CC/C=C/C=C/C=C/C=C/C(O[C@@H]2[C@@H](O)[C@@H](N)[C@H](O)[C@@H](C)O2)CC3O[C@](CC(O)C3C(O)=O)(O)CC(O)C(O)CCC(O)CC(O)CC(O)C1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27?,28?,29?,30?,31?,32?,33?,34?,35?,36?,37?,38?,40?,41-,42+,43+,44-,46+,47+/m0/s1
Formula Weight 925.5
Chemical Name or Material Nystatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.