missing translation for 'onlineSavingsMsg'
Learn More

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99%

Product Code. 11313097
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313097 25 g 25g
11393087 1 g 1g
11303097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313097 Supplier Thermo Scientific Alfa Aesar Supplier No. L13470.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 125700-67-6 | C11H16BF4N5O | 321.09 g/mol

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate is used as used for solid-phase peptide synthesis. It is also used to study ten different phosphonium and uronium salt-based coupling reagents, which are used to synthesize four peptides.

Solubility
Soluble in acetonitrile (100 mg/ml, clear, colorless), DMF (160.55 mg/ml, clear), and water (3 mg/ml at 20°C).

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2-8°C. Sensitive to light and moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125700-67-6
Molecular Formula C11H16BF4N5O
Molecular Weight (g/mol) 321.09
MDL Number MFCD00077413
InChI Key JKEKMBGUVUKMQB-UHFFFAOYSA-N
Synonym tbtu, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 2-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-1h-benzotriazol-1-yl-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-benzotriazol-1-yl uronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, o-benzotriazol-1-yl-n,n,n',n'-tetra methyluronium tetrafluoroborate
PubChem CID 2733207
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C12)=[N+](C)C

Specifications

Melting Point 195°C to 198°C (decomposition)
Quantity 25 g
Beilstein 7066325
Solubility Information Soluble in acetonitrile (100mg/ml,clear,colorless),DMF (160.55mg/ml,clear),and water (3mg/ml at 20°C).
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
Formula Weight 321.08
Percent Purity 99%
Chemical Name or Material O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.