missing translation for 'onlineSavingsMsg'
Learn More

O-(4-Nitrobenzyl)hydroxylamine hydrochloride, 98%

Product Code. 11387615
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387615 1 g 1g
11397615 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387615 Supplier Thermo Scientific Alfa Aesar Supplier No. L05861.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2086-26-2 | C7H9ClN2O3 | 204.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2086-26-2
Molecular Formula C7H9ClN2O3
Molecular Weight (g/mol) 204.61
MDL Number MFCD00012954
InChI Key LKCAFSOYOMFQSL-UHFFFAOYSA-N
Synonym o-4-nitrobenzyl hydroxylamine hydrochloride, 4-nitrobenzyloxyamine, o-4-nitrophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-4-nitrophenyl methyl-, monohydrochloride, p-nitrobenzyloxyamine hydrochloride, hydroxylamine, o-p-nitrobenzyl-, monohydrochloride, o-p-nitrobenzyl hydroxylamine hydrochloride, o-4-nitrobenzyl hydroxylammonium hydrochloride, 1-aminooxy methyl-4-nitrobenzene hydrochloride, hydroxylamine, o-p-nitrophenylmethyl-, monohydrochloride
PubChem CID 74967
IUPAC Name O-[(4-nitrophenyl)methyl]hydroxylamine;hydrochloride
SMILES [H+].[Cl-].NOCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∽215°C (decomposition)
Odor Nearly Odorless
Quantity 1 g
Beilstein 3709565
Sensitivity Light and moisture sensitive
Formula Weight 204.61
Percent Purity 98%
Chemical Name or Material O-(4-Nitrobenzyl)hydroxylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.