missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Acetyl-L-carnitine Hydrochloride

Product Code. 15484119
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484119 25 g 25g
15474119 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484119 Supplier Thermo Scientific Chemicals Supplier No. J61536.14
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

Weak cholinergic agonist | CAS: 5080-50-2 | C9H18ClNO4, C9H17NO4·HCl | 239.70 g/mol

O-Acetyl-L-carnitine hydrochloride is used as a cholinergic agonist that stimulates neuronal response to serotonin and acetylcholine. Acetylcarnitine has antinociceptive activity that may be mediated by enhanced activity of muscarinic cholinergic receptors or mGlu2 glutamate receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5080-50-2
Molecular Formula C9H17NO4·HCl, C9H18ClNO4
Synonym (R)-3-Acetoxy-4-(trimethylammonio)butyrate hydrochloride; L-Carnitine acetyl ester hydrochloride

Specifications

Quantity 25 g
Description White
Content And Storage 2°C to 8°C
For Use With (Application) Weak cholinergic agonist
Formula Weight 239.70g/mol
Sensitivity Hygroscopic
Product Type Signal Transduction Reagent
Form White crystalline powder
Beilstein 4340103
InChI Key JATPLOXBFFRHDN-UHFFFAOYNA-N
SMILES [H+].[Cl-].CC(=O)OC(CC([O-])=O)C[N+](C)(C)C
IUPAC Name hydrogen 3-(acetyloxy)-4-(trimethylazaniumyl)butanoate chloride
Molecular Weight (g/mol) 239.70
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.